MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates
Applied Biosystems™

MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates

MicroAmp EnduraPlate Optical 96-Well Clear Reaction Plates offer excellent PCR or qPCR performance to meet your experimental needs. EnduraPlate plasticRead more
Have Questions?
Catalog number A36924
Price (USD)/ 20 plates
165.65
Online exclusive
171.00 
Save 5.35 (3%)
-
Add to cart
Price (USD)/ 20 plates
165.65
Online exclusive
171.00 
Save 5.35 (3%)
Add to cart
MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates
Catalog numberA36924
Price (USD)/ 20 plates
165.65
Online exclusive
171.00 
Save 5.35 (3%)
-
Add to cart
MicroAmp EnduraPlate Optical 96-Well Clear Reaction Plates offer excellent PCR or qPCR performance to meet your experimental needs. EnduraPlate plastic consumables are validated with Applied Biosystems instruments for optimal fit and performance. They are constructed to ANSI/SBS standards and are well-suited for automated workflows. MicroAmp EnduraPlate Optical 96-Well Clear Reaction Plates are semi-skirted and fit standard (0.2 mL) 96-well thermal blocks.

• Optimized for performance with Applied Biosystems instruments
• Certified DNA-, RNase-, and PCR inhibitor-free
• Constructed to ANSI/SBS standards for compatibility with high-throughput laboratories and robotic handling
• Easy-to-read black well-identification text to minimize pipetting errors

EnduraPlate reaction plates use polypropylene for a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from the thermal block to the sample, while a polycarbonate frame provides mechanical stability, flatness, and durability that is compatible with automation/robotics. Easy-to-read, black well-identification text helps to minimize pipetting mistakes and improves sample preparation.

MicroAmp EnduraPlate Optical 96-Well Clear Reaction Plates are provided without barcodes, enabling customization with your own barcodes. For a barcoded version of this product, please see MicroAmp EnduraPlate Optical 96-Well Clear Reaction Plates with Barcode.

Specifications
BarcodeNo Barcode
Certifications/ComplianceCertified DNA/RNase Free
Packaging20 Plates
Product LineMicroAmp
ColorOptical, Clear
Product TypeMicroplate
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt StyleSemi
MaterialPolycarbonate, Polypropylene
Volume (Metric) Well0.3 mL
No. of Wells96
Volume (Metric) Working0.2 mL
Quantity20 plates
Shipping ConditionRoom Temperature
Unit Size20 plates
Contents & Storage
Store at room temperature.