High Select™ Depletion Spin Columns
High Select™ Depletion Spin Columns
Thermo Scientific™

High Select™ Depletion Spin Columns

Decrease albumin, immunoglobulins, and other high-abundance proteins from human serum or plasma in preparation for mass spectrometry analysis, or 1D and 2D gel electrophoresis using High Select depletion spin columns or resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
A36368HSA/Immunoglobulin Depletion Resin
A36365HSA/Immunoglobulin Depletion Mini Spin Columns
A36366HSA/Immunoglobulin Depletion Mini Spin Columns
A36367HSA/Immunoglobulin Depletion Midi Spin Columns
A36371Top14 Abundant Protein Depletion Midi Spin Columns
A36372Top14 Abundant Protein Depletion Resin
A36369Top14 Abundant Protein Depletion Mini Spin Columns
A36370Top14 Abundant Protein Depletion Mini Spin Columns
Catalog number A36368
Price (USD)/ 50 mL
2,718.65
Online exclusive
2910.00 
Save 191.35 (7%)
-
Add to cart
Description:
HSA/Immunoglobulin Depletion Resin
Price (USD)/ 50 mL
2,718.65
Online exclusive
2910.00 
Save 191.35 (7%)
Add to cart
High Select™ Depletion Spin Columns
Catalog numberA36368
Price (USD)/ 50 mL
2,718.65
Online exclusive
2910.00 
Save 191.35 (7%)
-
Add to cart
Analysis of human fluids is often complicated by the presence of high concentrations of serum proteins, including albumin and IgG that can make up more than 70% of total serum protein. Removal of albumin, immunoglobulins, and other abundant proteins is essential for the study of low-abundance proteins. Thermo Scientific High Select HSA/Immunoglobulin and Top 14 Abundant Protein depletion spin columns and resins enable depletion of the most abundant proteins in human serum and plasma for improved detection of low-abundance proteins using mass spectrometry analysis or 1D or 2D gel electrophoresis.
Flexible—resin provided in mini and midi column formats as well as bulk for custom filling options
Optimized—removes >95% of IgG and >95% of albumin (HSA/Immunoglobulin); removes >95% of IgG and >95% of albumin, plus 12 other abundant proteins (Top 14)
Fast—process samples in ∼10 minutes
Economical—cost-effective spin columns are priced for single use
Reduced carry-over—one-time use prevents protein carryover and experimental variability

High Select HSA/Immunoglobulin Depletion Resin uses highly specific, immobilized anti-HSA and anti-immuoglobulin antibodies (IgG, IgA, IgM, IgD, and IgE) to remove human serum albumin (HSA) and all major subclasses of immunoglobulins from serum and plasma. High Select Top14 Abundant Protein Depletion Resin uses highly specific immobilized antibodies to remove human serum albumin (HSA), albumin, IgG, IgA, IgM, IgD, IgE, kappa and lambda light chains, alpha-1-acidglycoprotein, alpha-1-antitrypsin, alpha-2-macroglobulin, apolipoprotein A1, fibrinogen, haptoglobin, and transferrin from serum, plasma, or spinal fluids. The resins are provided in an economical and convenient spin column format specifically designed for one-step processing and single use or bulk format for custom filling.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionHSA/Immunoglobulin Depletion Resin
For Use With (Equipment)Mass Spectrometer
Product TypeResin
Product LineHigh Select™
Label or DyeUnlabeled
Shipping ConditionWet Ice
Starting MaterialPlasma, Serum
Final Product TypeProtein
Workflow StepProtein Depletion
Quantity50 mL
Unit Size50 mL
Contents & Storage
Store at 2–8°C.