Albumin from Bovine Serum (BSA), Alexa Fluor™ 555 conjugate
This albumin from bovine serum (BSA), Alexa Fluor™ 555 conjugate is a bright and photostable conjugate exceptionally suited for applicationsRead more
Have Questions?
Catalog number A34786
Price (USD)/ 5 mg
320.65
Online exclusive
324.00
Save 3.35 (1%)
-
Add to cart
Quantity:
5 mg
Price (USD)/ 5 mg
320.65
Online exclusive
324.00
Save 3.35 (1%)
Add to cart
Albumin from Bovine Serum (BSA), Alexa Fluor™ 555 conjugate
Catalog numberA34786
Price (USD)/ 5 mg
320.65
Online exclusive
324.00
Save 3.35 (1%)
-
Add to cart
This albumin from bovine serum (BSA), Alexa Fluor™ 555 conjugate is a bright and photostable conjugate exceptionally suited for applications in which well-defined molecular weight tracers are needed. These applications include studying the rate of endocytosis and exocytosis and following intracellular protein processing.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity5 mg
ConjugateAlexa Fluor 555
SpeciesBovine
Label TypeAlexa Fluor Dyes
Shipping ConditionRoom Temperature
FormLyophilized
Protein SubtypeAlbumin Proteins
Product LineAlexa Fluor™
Unit Size5 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.