Fetal Bovine Serum, dialyzed, US origin
Fetal Bovine Serum, dialyzed, US origin
Gibco™

Fetal Bovine Serum, dialyzed, US origin

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A338200150 mLOne Shot
26400044500 mLBottle
Catalog number A3382001
Price (USD)/ 50 mL
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
50 mL
Format:
One Shot
Price (USD)/ 50 mL
116.65
Online exclusive
129.00 
Save 12.35 (10%)
Add to cart
Fetal Bovine Serum, dialyzed, US origin
Catalog numberA3382001
Price (USD)/ 50 mL
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
AgeFetal
SpeciesBovine
Country of OriginUnited States
FormLiquid
FormatOne Shot
Product TypeFetal Bovine Serum
Quantity50 mL
Serum TreatmentDialyzed
SterilitySterile
Product LineOne Shot™
Shipping ConditionFrozen
Unit Size50 mL
Contents & Storage
Storage conditions: ≤-10°C
Shipping conditions: Frozen