TRIzol™ Reagent and Phasemaker™ Tubes Complete System
Inquire about OEM or Commercial Supply version of this product here.
TRIzol™ Reagent and Phasemaker™ Tubes Complete System
Invitrogen™

TRIzol™ Reagent and Phasemaker™ Tubes Complete System

This system includes both TRIzol™ Reagent and Invitrogen™ Phasemaker™ Tubes. TRIzol Reagent is a complete, ready-to-use reagent for the isolationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A33251200 Preps
A33250100 Preps
Catalog number A33251
Price (USD)/ 200 preparations
-
Quantity:
200 Preps

This system includes both TRIzol™ Reagent and Invitrogen™ Phasemaker™ Tubes.

TRIzol Reagent is a complete, ready-to-use reagent for the isolation of high-quality total RNA from a variety of biological samples. It offers superior lysis capability, even with difficult sample types.

Phasemaker Tubes contain a thick liquid polymer that separates and completely isolates the upper aqueous phase of the TRIzol Reagent mix from the organic phase underneath, greatly simplifying removal of the upper phase containing the RNA. This TRIzol Reagent and Phasemaker Tubes Complete System is for the extraction of RNA, not DNA or protein. It will accommodate 50–100 mg of tissue. For cultured cells, please refer to the manual for information on sample size.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Isolation TechnologyOrganic Extraction
Purification Time1 hr.
Elution Volume20 to 50 μL
Sample TypeBacteria, Cells, Plants, Tissue, Yeast
Final Product TypeTotal RNA
High-throughput CompatibilityNot High-throughput Compatible (Manual)
For Use With (Application)RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
Quantity200 Preps
Shipping ConditionRoom Temperature
Starting Material AmountBacteria: ≤107 cells
Cells: ≤107
Plant: ≤107 cells, 100 mg
Tissue: ≤100 mg
Yeast: ≤107 cells
YieldCells: ≤15 μg
Tissue: ≤73 μg
Unit Size200 preparations
Contents & Storage
• 200 mL TRIzol Reagent; room temperature
• 200 Phasemaker Tubes; room temperature