POROS™ XQ RoboColumn™ Pre-packed columns
Product Image
Thermo Scientific™
POROS™ XQ RoboColumn™ Pre-packed columns
Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColumn Capacity
A30720600 μL
A30719200 μL
Catalog number A30720
Price (USD)/ 600 µL
341.00
-
Add to cart
Column Capacity:
600 μL
Price (USD)/ 600 µL
341.00
Add to cart
POROS™ XQ RoboColumn™ Pre-packed columns
Catalog numberA30720
Price (USD)/ 600 µL
341.00
-
Add to cart
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS XQ resin, a 50-micron strong anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that that enables >140 mg/mL dynamic binding capacity in the presence of up to 6.6 mS/cm NaCl, while delivering exceptional separation performance. A major improvement to the performance of traditional anion exchangers, POROS XQ resin sets the standard for strong anion exchange chromatography in both bind and elute or flow-through modes of operation.
Features of the resin:
  • High dynamic binding capacity leading to reduced column size, smaller footprint, decreased water and buffer usage, and reduced cycling
  • High resolution supporting improved product yield and purity, ultimately improving the cost of goods for the molecule being produced
  • Robust salt tolerance allowing enhanced process design flexibility and streamlining of process flow
  • Low operating back pressure and linear pressure versus flow responses driving flexible scalability
  • Rigid polymeric bead with covalent surface chemistry for easier handling and packing, and superior physical and chemical stability enabling a more robust downstream purification process
RoboColumn™ is a registered trademark of ATOLL GmbH.
Specifications
Column Capacity600 μL
Flow Rate16 to 1000 cm/hr.
Unit Size600 µL
Contents & Storage
Store at 2°C to 8°C or at room temperature.