Alexa Fluor™ Plus 750 Phalloidin
Alexa Fluor™ Plus 750 Phalloidin
Invitrogen™

Alexa Fluor™ Plus 750 Phalloidin

Alexa Fluor Plus 750 Phalloidin is a high-affinity filamentous actin (F-actin) probe (phalloidin) conjugated to our bright, photostable, near-IR AlexaRead more
Have Questions?
Catalog number A30105
Price (USD)/ 1 vial
597.65
Online exclusive
636.00 
Save 38.35 (6%)
-
Add to cart
Price (USD)/ 1 vial
597.65
Online exclusive
636.00 
Save 38.35 (6%)
Add to cart
Alexa Fluor™ Plus 750 Phalloidin
Catalog numberA30105
Price (USD)/ 1 vial
597.65
Online exclusive
636.00 
Save 38.35 (6%)
-
Add to cart
Alexa Fluor Plus 750 Phalloidin is a high-affinity filamentous actin (F-actin) probe (phalloidin) conjugated to our bright, photostable, near-IR Alexa Fluor Plus 750 dye.

• Selectively stains F-actin
• Outstanding fluorescence performance
• Excitation/emission: 758/784 nm. Compatible with standard Cy7 filter set.
• Superior to antibody staining
• Optimal for fixed and permeabilized samples

Get superior results in your actin staining studies
Phalloidin is a bicyclic peptide belonging to a family of toxins isolated from the deadly Amanita phalloides ''death cap'' mushroom and commonly used in imaging applications to selectively label F-actin. Fluorescently-labeled phalloidin has several advantages over antibodies for actin labeling, including virtually identical binding properties with actin from different species of plants and animals, and low non-specific binding. Phalloidin binds F-actin with high selectivity, while Alexa Fluor Plus 750 provides violet fluorescence of unparalleled brightness and photostability.

Use in multiple applications
Alexa Fluor 750 Plus Phalloidin can be used to visualize and quantitate F-actin in tissue sections, cell cultures, or cell-free preparations. Its staining is fully compatible with other fluorescent stains used in cellular analyses, including fluorescent proteins, Qdot nanocrystals, and other Alexa Fluor conjugates including secondary antibodies.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Excitation Wavelength Range758/784
Dye TypeAlexa Fluor™ Plus 750
Quantity1 vial
Product LineAlexa Fluor™ Plus
Shipping ConditionQualified for Ambient or Wet Ice
Label TypeAlexa Fluor Dyes
Product TypePhalloidin
SubCellular LocalizationActin
Unit Size1 vial
Contents & Storage
1 vial sufficient for 300 units/slides
Store at ≤-20°C after receiving
Desiccate, and protect from light
When stored as directed, product is stable for at least 1 year.