Attune™ NxT External Fluid Supply
Product Image
Invitrogen™
Attune™ NxT External Fluid Supply
The Attune™ NxT External Fluid Supply is an accessory for the Attune™ NxT Acoustic Focusing Cytometer. It was designed toRead more
Have Questions?
Catalog number A28006
Price (USD)
-
Request A Quote
The Attune™ NxT External Fluid Supply is an accessory for the Attune™ NxT Acoustic Focusing Cytometer. It was designed to facilitate the use of a larger buffer and waste container. The system works by monitoring and supplying the internal fluid tanks on the Attune NxT instrument so that the system can run without having to stop instrument operation to change out the fluid tanks or empty the waste. The external fluid system uses the 10 L cube of Attune™ Focusing Fluid (Cat. No. A24904) and a Nalgene™ waste bottle. The external fluid supply is ideal for use with the Attune™ NxT Autosampler and enables the collection of approximately 28 96-well or 7 384-well plates without changing the fluid tanks.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Attune™ NxT Acoustic Focusing Cytometer
Product LineAttune™
Shipping ConditionRoom Temperature
Product TypeFlow Cytometer Accessory
Unit Size