RapidFinder™ Quant Multi-Animal Set
RapidFinder™ Quant Multi-Animal Set
Thermo Scientific™

RapidFinder™ Quant Multi-Animal Set

The RapidFinder™ Quant Multi-Animal Set, when used with one or more of the RapidFinder™ Animal Species ID kits listed below,Read more
Have Questions?
Catalog number A24399
Price (USD)/ 48 reactions
972.00
-
Add to cart
Price (USD)/ 48 reactions
972.00
Add to cart
RapidFinder™ Quant Multi-Animal Set
Catalog numberA24399
Price (USD)/ 48 reactions
972.00
-
Add to cart
The RapidFinder™ Quant Multi-Animal Set, when used with one or more of the RapidFinder™ Animal Species ID kits listed below, enables the determination of the percentage of a target animal species with respect to total animal in food and feed samples. Using the included standards, the kit enables quantification of as little as 0.05% target species DNA as compared to other animal DNA in only a few hours.

The RapidFinder Quant Multi-Animal Set is designed to be used in combination with one or more of the following animal species-specific ID kits (available separately):

Name Catalog Number
RapidFinder™ Beef ID Kit A24391
RapidFinder™ Pork ID Kit A24392
RapidFinder™ Equine ID Kit A15570
RapidFinder™ Poultry ID Kit A24397
RapidFinder™ Chicken ID Kit A24393
RapidFinder™ Turkey ID Kit A24394
RapidFinder™ Halal ID Kit A47086
RapidFinder™ Sheep ID Kit A24395
RapidFinder™ Goat ID Kit A24407
RapidFinder™ Fallow Deer ID Kit A24410


• Simple-real-time PCR based assay, no electrophoresis needed
• Flexible-compatible with any food or feed sample, both raw and processed
• Reliable-Includes multi-animal DNA quantification standard
• Fast-from sample to result in the same day

How It Works
The method relies on quantitative real-time PCR. First, a standard curve is created by serially diluting the included multi-animal DNA standard. Then assays for the target species DNA (from one of the kits listed above) and for all animal DNA (from the RapidFinder™ Quant Multi-Animal Set) are run side-by-side on both the food sample DNA and the DNA standards. This approach enables rapid and accurate quantification and reporting of the percentage of target animal species as compared to total animal in a sample. Included in each reaction mix is an internal positive control detected in the VIC™ dye channel, which is used to verify the absence of PCR inhibitors.

The RapidFinder™ Quant Multi-Animal Set is optimized and validated for use with the Applied Biosystems™ QuantStudio™ 5 Real‑Time PCR Instrument and the Applied Biosystems™ 7500 Fast Real‑Time PCR Instrument.

Sample Preparation
DNA sample preparation for the RapidFinder™ animal species identification and quantification workflow is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using 10 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.
Specifications
Detection MethodPrimer-probe
PCR MethodqPCR
Reaction SpeedStandard
For Use With (Equipment)Applied Biosystems™ QuantStudio™ 5 Real‑Time PCR Instrument and Applied Biosystems™ 7500 Fast Real‑Time PCR Instrument
Product LineRapidFinder™
SpeciesMulti-animal
For Use With (Application)Food and Beverage
No. of Reactions48 reactions
Sample TypeFood & Environmental Samples
Unit Size48 reactions
Contents & Storage
Multi-Animal Master Mix (store at -20°C), General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use. Store protected from light.), Multi-Animal Standard (store at -20°C)