PolarScreen™ ER Beta Competitor Assay Kit, Green
PolarScreen™ ER Beta Competitor Assay Kit, Green
Thermo Scientific™

PolarScreen™ ER Beta Competitor Assay Kit, Green

The PolarScreen™ ER Beta Competitor Assay Kit provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potentialRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A158892500 Assays
A15890800 Assays
Catalog number A15889
Price (USD)/ 2,500 assays
2,950.00
-
Add to cart
Quantity:
2500 Assays
Price (USD)/ 2,500 assays
2,950.00
Add to cart
PolarScreen™ ER Beta Competitor Assay Kit, Green
Catalog numberA15889
Price (USD)/ 2,500 assays
2,950.00
-
Add to cart

The PolarScreen™ ER Beta Competitor Assay Kit provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential estrogen receptor (ER) beta ligands. The kit uses insect cell-expressed, full-length, native (untagged) human ER beta (Cat. No. A15664) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone™ ES2) in a homogenous, mix-and-read assay format.

This kit produces a green color read-out; a 'red' kit is also available (A15891). Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance email drugdiscoverytech@lifetech.com.

The PolarScreen™ ER Beta Competitor Assay Kit is:

  • Complete & ready-to-use out of the box—all reagents included, just add your test compounds
  • High throughput compatible—20 μL fluorescence polarization assays in a 384-well plate
  • Convenient—available in bulk sizes for larger screening needs

How the Assay Works

Full-length ER beta is added to a fluorescent estrogen ligand to form an ER-Fluormone™ ES2 complex. This complex is added to individual test compounds in multiwell plates. To determine relative affinity, the assay measures fluorescence polarization. Competing test compounds will displace the Fluormone™ ES2 ligand from ER beta, permitting it to tumble rapidly and resulting in low polarization values. A shift in the fluorescence polarization value in the presence of test compound is used to determine the relative affinity of test compounds for ER beta.

All components are manufactured under the strictest quality parameters. This product's performance, per the Certificat of Analysis, is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Quantity2500 Assays
Packaging384-well Plate
Assay CategoryBiochemical Assay
Druggable TargetNuclear Receptors
ReadoutFluorescence Polarization
Product LinePolarScreen™
For Use With (Application)Nuclear Receptor Assay, Fluorescence Polarization
LigandER beta
Product TypeAssay Kit
Target MoleculeEstrogen Receptor Beta Nuclear Receptor
Unit Size2,500 assays
Contents & Storage
• Estrogen Receptor (ER) Beta Recombinant Protein, Full Length (store at -68°C to -85°C)
• Fluromone™ ES2 (store at -5°C to -30° C)
• ES2 Screening Buffer (store at room temperature)