PBS, pH 7.4 (flow cytometry grade)
Save up to 31% on your order of cell culture products. View promotion details »
PBS, pH 7.4 (flow cytometry grade)
Gibco™

PBS, pH 7.4 (flow cytometry grade)

Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of applications, including reagent preparation, diluting cells forRead more
Have Questions?
Catalog number A1286301
Price (USD)/ 10 L
82.65
Online exclusive
84.00 
Save 1.35 (2%)
-
Add to cart
Price (USD)/ 10 L
82.65
Online exclusive
84.00 
Save 1.35 (2%)
Add to cart
PBS, pH 7.4 (flow cytometry grade)
Catalog numberA1286301
Price (USD)/ 10 L
82.65
Online exclusive
84.00 
Save 1.35 (2%)
-
Add to cart

Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of applications, including reagent preparation, diluting cells for flow cytometry and as a cell culture reagent. This Gibco™ PBS is formulated without calcium and magnesium for suspending cells for flow cytometry. This PBS is not intended for cell culture as it is not tested for sterility. We offer a variety of Gibco™ PBS formulations for cell culture applications. Find the right cell culture product using the media selector tool.

This PBS is manufactured as follows:

Without
• Calcium
• Magnesium
• Phenol Red


Product Intended Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AdditiveNo Calcium, No Magnesium
Dilution1 X
Product TypePhosphate Buffered Saline (PBS)
Quantity10 L
Solution TypePhosphate Buffered Saline
pH7.4
Phenol Red IndicatorNo Phenol Red
Shipping ConditionRoom Temperature
SterilitySterile-filtered
FormLiquid
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Unit Size10 L
Contents & Storage
Storage conditions: 15–30°C
Shipping conditions: Ambient
Shelf life: 9 months from date of manufacture