Gallery Reagents for Food and Beverage Analysis
Gallery Reagents for Food and Beverage Analysis
Actual product may vary
Thermo Scientific™
Gallery Reagents for Food and Beverage Analysis
Choose from a continuously expanding selection of tests for automated food and beverage analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
984348Oxalic Acid system reagents (250 tests)
984300Ethanol system reagents (300 tests)
984302D-Fructose system reagents (300 tests)
981842Wash Fluid
984304D-Glucose system reagents (300 tests)
984332ECM pH 7 Standard tests
984353BC System liquid system reagents (360 tests)
984354BC Diluent system reagents (360 tests)
984355BC Eluent system reagents (360 tests)
984764D-Glucose (1000 tests)
984765Glycerol system reagents (1000 tests)
984305Beta-Glucan (High MW) system reagents (350 tests)
984306D-Lactic Acid system reagents (300 tests)
984307Potassium system reagents (300 tests)
984308L-Lactic Acid system reagents (300 tests)
984309Tartaric Acid system reagents (480 tests)
984310L-Malic Acid system reagents (300 tests)
984312Sucrose (Total Glucose) system reagents (300 tests)
984314D-Glucose + D-Fructose system reagents (300 tests)
984315Lipase system reagents (500 tests)
984316Glycerol system reagents (300 tests)
984317D-Glucose + D-Fructose + Sucrose system reagents (300 tests)
984318Acetic Acid system reagents (300 tests)
984319L-Asparagine/L-Aspartic Acid (350 tests)
984320Ammonia system reagents (300 tests)
984321Urea (Ammonia) system reagents (300 tests)
984322D-Isocitric Acid system reagents (300 tests)
984323Lactose (Glucose) system reagents (300 tests)
984324Cholesterol in Food system reagents (300 tests)
984326Total Iron (Fe) system reagents (850 tests)
984327Citric Acid system reagents (250 tests)
984328Total Protein (Biuret) system reagents (450 tests)
984342Alpha-Amino Nitrogen by NOPA system reagents (300 tests)
984345Total Sulfite (SO2) system reagents (300 tests)
984346Total Polyphenol system reagents (280 tests)
984347Acetaldehyde system reagents (250 tests)
984349pH (colorimetric) system reagents (330 tests)
984356Acetic Acid by ACS method system reagents (150 tests)
984380Sugar Combination Standard for D-Fructose (0.500 g/L), D-Glucose (0.500 g/L), Sucrose (0.500 g/L), or total Glucose (0.760 g/L) tests
984382Acid Combination Standard for D-Lactic acid (0.200 g/L), L-Malic acid (0.500 g/L), L-Lactic acid (220 mg/L), and D-Lactic acid (220 mg/L) tests
984383Beta-Glucan standard for Beta-Glucan (High MW) test (500 mg/L)
984384Ethanol standard reagent (0.5 g/L)
984386Glycerol standard reagent (0.21 g/L)
984388Urea standard reagent (500 mg/L)
984390Lactose standard for Lactose (Glucose) tests (0.95 g/L)
984391Cholesterol standard for Cholesterol in Food tests (2000 mg/L)
984393Oxalic Acid standard (45 mg/L)
984394NOPA Standard for Alpha-Amino Nitrogent test (150 mg/L)
984396Acetaldehyde Standard (100 mg/L)
984632Total Acids (Wine pH 7) system reagents (400 tests)
984633Total acids (Juice pH 8) system reagents (400 tests)
984634Free sulfite (SO2) system reagents (430 tests)
984635L-Ascorbic Acid system reagents (250 tests)
984636L-Glutamic Acid system reagents (300 tests)
984710D-Gluconic Acid system reagents (250 tests)
984373Alpha-Amylase system reagents (200 tests)
Catalog number 984348
Price (USD)/ Each
-
Request A Quote
Description:
Oxalic Acid system reagents (250 tests)
Save time by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for food and beverage analysis. Over 50 ready-to-use gallery system reagents are offered for food and beverage analysis, with the kits including two to four reagents in 20 mL barcoded vials. This enables the necessary identification data, such as lot numbers and expiration dates, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations. Save time and reduce errors by simplifying your analysis with the Gallery system reagents.
  • Kit structure includes 2 to 4 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, is automatically read
  • Photometric measurement is mainly based on the formation or consumption of NAD or NADH
  • Measurements are done at 340nm in 37°C
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
Specifications
TypeOxalic Acid System Reagent
Sample TypeFood and Beverage Sample
DescriptionOxalic Acid system reagents (250 tests)
DetectsOxalic Acid
No. of Tests250
Test Range2 to 100 mg/L
Analytical MethodEnzymatic test with oxalate oxidase
Quantity2 Reagents in 20 mL Vials
Interference Filter600/ 700
Unit SizeEach