GFS33 Graphite Furnace and Autosampler for iCE™ 3300 AA spectrometers
GFS33 Graphite Furnace and Autosampler for iCE™ 3300 AA spectrometers
Actual product may vary
Thermo Scientific™
GFS33 Graphite Furnace and Autosampler for iCE™ 3300 AA spectrometers
Provide the ideal combination of AA flame and furnace technology for superb laboratory adaptability with the Thermo Scientific™ GFS33 Graphite Furnace and Autosampler for iCE™ 3300 AA spectrometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
942359030001GFS33 Integrated Graphite Furnace and Autosampler; For iCE 3300
942359030002GFS33 Integrated Graphite Furnace and Autosampler; For iCE 3300 (North American Version)
Catalog number 942359030001
Price (USD)/ Each
-
Request A Quote
Description:
GFS33 Integrated Graphite Furnace and Autosampler; For iCE 3300
Provide a combination of flame and furnace technology that offer users unparalleled AA instrument sensitivity and productivity with a single module, combined graphite furnace and autosampler. Using the Thermo Scientific™ GFS33 Graphite Furnace and Autosampler for iCE™ 3300 AA spectrometer gives flexibility and adaptability to the routine laboratory.
Increase Sensitivity and Save Time
Flame to furnace changeover is a simple routine operation on our single atomizer iCE 3300 AA system, as the burner does not have to be removed. This combined, pre-aligned GFS33 Graphite Furnace and Autosampler module simplifies installation and eliminates the need for daily optimization.
  • Dynamic, optical cuvette temperature control and pre-heated cuvette injection
  • Thermo Scientific™ Extended Lifetime Cuvettes (ELC) provide uninterrupted overnight analysis and lower cost of ownership
  • QuadLine™, high speed, high energy deuterium source correction, offers maximum sensitivity and wider working range
  • Corrects up to 2A of background, with less than 2% residual error even for fast transient signals
  • Autosampler prepares calibration standards, adds matrix modifiers, and dilutes samples
  • Dilutes over-range samples during an analysis
  • Software functionality to use the method of standard additions and perform automated Ash-Atomize experiments
  • Compatible with the iCE 3300 AA systems
Specifications
TypeIntegrated Graphite Furnace and Autosampler
DescriptionGFS33 Integrated Graphite Furnace and Autosampler; For iCE 3300
For Use With (Equipment)iCE 3300 AA Spectrometers
Unit SizeEach