Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Actual product may vary
Thermo Scientific™

Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantity
941709Complextion Ionic Strength Adjuster (CISA) for Chloride Electrode2 x 1 L
841101Storage Solution for Sodium Electrode475 mL
841111Ionic Strength Adjustor Buffer Solution for Sodium Ion Selective Electrodes, 475mL475 mL
841113Reconditioning Solution for Sodium Electrode475 mL
930710Nitrate Interference Suppressor Solution for Nitrate Electrode475 mL
930711ISA for Fluoroborate, Nitrate, Perchlorate Electrodes475 mL
931911ISA for Potassium Electrode475 mL
932011ISA for Calcium Electrode475 mL
934610ISA for Nitrite Electrode475 mL
940011ISA for Bromide, Cadmium, Chloride, Cupric, Iodide, Sufide, Thiocyanate Electrodes475 mL
940909TISAB II for Fluoride Electrode1 Gal.
940911TISAB III Concentrate for Fluoride Electrode475 mL
940999TISAB II for Fluoride Electrode4 x 1 Gal.
941609Sulfide Anti-Oxidant Buffer (SAOB) for Sulfide Electrode4 x 475 mL
950210ISA for Carbon Dioxide Electrode475 mL
951011Alkaline Reagent for Cyanide Electrode475 mL
951210pH Adjusting Low Level ISA for Ammonia Electrode475 mL
951211pH Adjusting ISA for Ammonia Electrode475 mL
951213Storage Solution for Ammonia Electrode475 mL
956410Acid Buffer for Nitrogen Oxide Electrode475 mL
977010Iodide Reagent for Residual Chlorine Electrode5 x 50 mL
977011Acid Reagent for Residual Chlorine Electrode475 mL
Catalog number 941709
Price (USD)/ Pack of 2
340.29
Special Offer
Online exclusive
Ends: 31-Dec-2024
398.00 
Save 57.71 (15%)
-
Add to cart
Description:
Complextion Ionic Strength Adjuster (CISA) for Chloride Electrode
Quantity:
2 x 1 L
Price (USD)/ Pack of 2
340.29
Special Offer
Online exclusive
Ends: 31-Dec-2024
398.00 
Save 57.71 (15%)
Add to cart
Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Catalog number941709
Price (USD)/ Pack of 2
340.29
Special Offer
Online exclusive
Ends: 31-Dec-2024
398.00 
Save 57.71 (15%)
-
Add to cart

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.

Warranty: Out of box

Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements. The ammonia pH-adjusting ISA (951211) and low-level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low-level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.

All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:

  • Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
  • Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
  • Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
  • Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Concentration10 ppm
DescriptionComplextion Ionic Strength Adjuster (CISA) for Chloride Electrode
For Use With (Equipment)Ion Selective Electrode
TypeIon Selective Electrode Reagents
For Use With (Application)Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
Quantity2 x 1 L
Unit SizePack of 2