Make the most of the power of nuclear magnetic resonance (NMR) spectroscopy with the compact, affordable Thermo Scientific™ picoSpin™ 45 Series II NMR Spectrometer.
Get a dedicated NMR spectrometer for your research, manufacturing process or classroom with the affordable Thermo Scientific™ picoSpin™ 45 Series II NMR spectrometer. A true breakthrough in chemical instrumentation, the picoSpin 45 NMR Spectrometer occupies a fraction of the space of a conventional NMR spectrometer and doesn't require specialized knowledge or training for operation. Liquid samples are simply injected into an internal capillary via front-panel fittings, and only 30 microliters of sample fluid is needed to obtain a spectrum.
The picoSpin 45 Series II NMR spectrometer offers simplified workflows, a modern interface and automatic shimming plus custom methods; perfect for novice or experienced users alike.
Designed for ease of use, liquid samples are simply injected into an internal capillary via front-panel fittings and only 30 microliters of sample fluid is needed to obtain a spectrum. A highly stable, temperature-controlled permanent magnet ensures easy maintenance-free operation without the need for liquid cryogens. NMR data is generated in the industry-standard JCAMP-DX format for compatibility with standard NMR data analysis packages. The unit's fluid capillary is contained within a cartridge, so it can be replaced easily by the user in case it becomes blocked or contaminated.
Capillary Sample Loop, Capillary ID: 400 μm, Sample Volume: 30 μL
1/16 in. Interface Fittings
User Replaceable