Orion™ pH Buffer Cubitainers™
Orion™ pH Buffer Cubitainers™
Actual product may vary
Thermo Scientific™

Orion™ pH Buffer Cubitainers™

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpHPackagingQuantity
9110CB10.01Cubitainer19 L
9104CB4.01Cubitainer19 L
9107CB7Cubitainer19 L
Catalog number 9110CB
Price (USD)/ Each
167.70
Online exclusive
195.00 
Save 27.30 (14%)
-
Add to cart
pH:
10.01
Packaging:
Cubitainer
Quantity:
19 L
Price (USD)/ Each
167.70
Online exclusive
195.00 
Save 27.30 (14%)
Add to cart
Orion™ pH Buffer Cubitainers™
Catalog number9110CB
Price (USD)/ Each
167.70
Online exclusive
195.00 
Save 27.30 (14%)
-
Add to cart

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.

  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST™ traceability
  • Manufactured under ISO 9000 quality standards
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Chemical Name or MaterialpH Buffer, 5 gallon cubitainers
Boiling Point100°C
ColorLight Blue
DescriptionpH 10.01 Buffer, 5 gallon
For Use With (Application)Accurate, reproducible pH calibrations
For Use With (Equipment)pH Meter and Electrode
pH10.01
Physical FormLiquid
Quantity19 L
FormulationWater, Sodium Bicarbonate, Methylparaben, FD & C Blue #1, Sodium Carbonate
PackagingCubitainer
Traceability to NISTYes
Solubility InformationSoluble
Unit SizeEach