Achieve highly efficient, compatible immunoprecipitation and co-immunoprecipitation of target antigens for downstream LC-MS analysis using Thermo Scientific Pierce MS-Compatible Magnetic IP Kits with streptavidin or Protein A/G magnetic beads.
Have Questions?
Change view
Catalog Number
Description
90408
Streptavidin Magnetic Beads
90409
Protein A/G Magnetic Beads
Catalog number 90408
Price (USD)/ 40 reactions
558.65
Online exclusive
590.00
Save 31.35 (5%)
-
Add to cart
Description:
Streptavidin Magnetic Beads
Request bulk or custom format
Price (USD)/ 40 reactions
558.65
Online exclusive
590.00
Save 31.35 (5%)
Add to cart
Pierce™ MS-Compatible Magnetic IP Kits
Catalog number90408
Price (USD)/ 40 reactions
558.65
Online exclusive
590.00
Save 31.35 (5%)
-
Add to cart
Immunoprecipitation is often necessary for the detection and quantitation of low-abundance proteins or rare signaling events. Mass spectrometry analysis relies on two methods for the detection of low-abundance proteins: single reaction monitoring (SRM) and parallel reaction monitoring (PRM), which enable isolation, scheduling, and quantitation of selected proteotypic peptides. These IP kits have been optimized for efficient immunoprecipitation and co-immunoprecipitation of target proteins, and are directly compatible with downstream MS sample preparation and analysis.
• MS-compatible—reagents directly compatible with in-solution peptide digestion • Sensitive—procedure successfully enriches low-abundance proteins (low ng range) • Low background—enriched samples contain minimal detergent residuals • Reduced antibody contamination—antibody biotinylation with streptavidin enrichment reduces contamination in the eluate when compared to other IP methods • Robust—procedure and reagents have been verified with numerous targets to ensure consistent enrichment of low-abundance proteins (ng range) with at least two peptides identified per protein
The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Protein A/G) uses high-quality Pierce Protein A/G Magnetic Beads to provide wider flexibility of antibody capture than either Protein A or G alone. The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Streptavidin) uses high-quality Pierce Streptavidin Magnetic Beads for use with biotinylated antibodies that may be purchased commercially or labeled using the Pierce Antibody Biotinylation Kit for IP. After the immunoprecipitation procedure, the target-enriched elution fraction is ready for in-solution tryptic digestion for LC-MS analysis without the need for gel purification, detergent removal, or desalting. Additionally, the reagents and procedures have been validated using both manual and automated magnetic separation.
Applications • Enrichment of low-abundance targets to improve MS sensitivity • Identification of target proteins that are not detectable by western blotting • Quantitation of target proteins by LC-MS analysis • Screening of antibodies for IP-MS
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionStreptavidin Magnetic Beads
For Use With (Equipment)KingFisher™ Flex, Magnetic Stand
Product TypeMagnetic IP Kit
Product LinePierce™
Sufficient For40 Reactions
Starting MaterialProtein Samples, Cell Lysate
Final Product TypeProtein
Detection MethodMass Spectrometry
FormatKit
Workflow StepSample Enrichment
Quantity1 Kit
For Use With (Application)Mass Spectrometry
Unit Size40 reactions
Contents & Storage
Streptavidin Magnetic Beads, 1 mL IP-MS Cell Lysis Buffer, 100 mL IP-MS Wash Buffer A, 75 mL IP-MS Wash Buffer B, 40 mL IP-MS Elution Buffer, 6 mL