Axiom™ GeneTitan™ Consumables Kit
Axiom™ GeneTitan™ Consumables Kit
Applied Biosystems™

Axiom™ GeneTitan™ Consumables Kit

Axiom™ GeneTitan™ Consumables Kit includes all GeneTitan instrument consumables required to process one Axiom 96 array plate.Read more
Have Questions?
Catalog number 901606
Price (USD)/ 1 plate
-
Axiom™ GeneTitan™ Consumables Kit includes all GeneTitan instrument consumables required to process one Axiom 96 array plate.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeConsumables Kit
Format96-array Plate
Quantity1 plate
Reagent TypeGenetitan consumables
Unit Size1 plate
Contents & Storage
Store all at room temperature
Contains:
5 X 96 layout stain trays
1 X 96 layout Hybridization tray
1 X 96 layout Scan tray
6 X 96 layout Scan and Stain tray covers