Pierce™ Protein A/G Chromatography Cartridges
Pierce™ Protein A/G Chromatography Cartridges
Actual product may vary
Thermo Scientific™

Pierce™ Protein A/G Chromatography Cartridges

The Thermo Scientific™ Pierce™ Chromatography Cartridges Protein A/G are convenient, ready-to-use prepacked devices for isolation and purification of most immunoglobins.PierceRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantity
89931Pierce Protein A/G Chromatography Cartridges, 5mL1 Cartridge
89930Pierce Protein A/G Chromatography Cartridges, 1mL2 Cartridges
Catalog number 89931
Price (USD)/ 1 x 5 mL
652.00
-
Add to cart
Description:
Pierce Protein A/G Chromatography Cartridges, 5mL
Quantity:
1 Cartridge
Request bulk or custom format
Price (USD)/ 1 x 5 mL
652.00
Add to cart
Pierce™ Protein A/G Chromatography Cartridges
Catalog number89931
Price (USD)/ 1 x 5 mL
652.00
-
Add to cart
The Thermo Scientific™ Pierce™ Chromatography Cartridges Protein A/G are convenient, ready-to-use prepacked devices for isolation and purification of most immunoglobins.

Pierce Protein A/G Agarose consists of purified Protein A/G recombinant fusion protein that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose (CL-6B). This particular variety of the resin provides the most versatile combination of chromatographic features for high yield and high purity purification of whole IgG from mammalian serum samples. The agarose beads have physical and chemical properties suitable for many affinity purification systems.

Features of Protein A/G Agarose:

Protein A/G – immobilized recombinant fusion protein of the antibody-binding domains of Protein A and Protein G enables polyclonal IgG purification from nearly any mammalian species
Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
Inert and stable – superior manufacturing method immobilizes Protein A/G by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
Standard capacity – Pierce Protein A/G Agarose has a normal load of immobilized Protein A/G, providing a binding capacity greater than 7 mg human IgG/mL resin

Pierce Chromatography Cartridges are compatible with the major automated liquid-chromatography systems or for manual syringe processing, and attach directly to ÄKTA™ or FPLC Systems without additional connectors. An accessory pack, included with each product, readily adapts cartridges for use with Luer-Lok Syringe Fittings or 1/16” tubing. The Protein A/G cartridges provide fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.

Protein A/G is a genetically-engineered protein that combines the IgG binding domains of both Protein A and Protein G. The fusion protein is expressed in E. coli. Protein A/G contains four Fc binding domains from Protein A and two from Protein G, resulting in a final mass of 50,460 daltons (40 to 45kDa by SDS-PAGE). Protein A/G is not as pH-dependent as Protein A alone, but otherwise has the additive properties of Protein A and G.

Protein A/G binds to all human IgG subclasses, making it the ideal choice for purification of polyclonal or monoclonal IgG antibodies whose subclass identities have not been determined. In addition, it binds to IgA, IgE, IgM and (to a lesser extent) IgD. Protein A/G also binds well to all mouse IgG subclasses but does not bind mouse IgA, IgM or serum albumin. This makes Protein A/G an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses, without interference from IgA, IgM and murine serum albumin. Individual subclasses of mouse monoclonals are likely to have a stronger affinity to the chimeric Protein A/G than to either Protein A or Protein G.

Pierce Protein A/G Agarose is prepared using Thermo Scientific AminoLink Coupling Chemistry, which provides several advantages compared to traditional methods of ligand immobilization. AminoLink Immobilization results in conjugation between sugar monomers of the agarose beads and native lysine residues on the Protein A via simple amide bonds. Unlike typical cyanogen bromide (CNBr) immobilization, the AminoLink Method does not introduce any novel chemical groups that could cause undesired nonspecific binding and produces a stable, essentially irreversible bond. The result is a high-binding-capacity resin that retains functional immobilized Protein A/G through multiple rounds of antibody purification.

Pierce Protein A/G Agarose binds nearly all isotypes and mammalian species of IgG from serum, ascites fluid, cell culture supernantant and other ant
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeChromatography Cartridge
Product LinePierce™
Column TypeAgarose Resin, Affinity
FormatChromatography Cartridge
Stationary PhaseProtein A/G
Purification TargetIgG Antibodies
DescriptionPierce Protein A/G Chromatography Cartridges, 5mL
Quantity1 Cartridge
Unit Size1 x 5 mL
Contents & Storage
Upon receipt store at 4°C to 8°C. Do not freeze.