Touch Screen Dry Bath/Block Heater
Touch Screen Dry Bath/Block Heater
Thermo Scientific™

Touch Screen Dry Bath/Block Heater

Achieve precise temperature stability and uniformity with our easy-to-use touch screen dry bath.
Have Questions?
Change viewbuttonViewtableView
No. of Blocks:
4 (must be ordered separately)
1 (order separately)
2 (must be ordered separately)
Voltage:
100/120 V
200/240 V
Includes:
View all
Dry bath, US style plug, block lifter
Heating Rate:
View all
≤ 25 min. 30° to 130°C
Catalog number 88870009
Price (USD)/ Each
1,098.00
Online exclusive
1220.00 
Save 122.00 (10%)
-
Add to cart
No. of Blocks:
4 (must be ordered separately)
Voltage:
100/120 V
Includes:
Dry bath, US style plug, block lifter
Heating Rate:
≤ 25 min. 30° to 130°C
Price (USD)/ Each
1,098.00
Online exclusive
1220.00 
Save 122.00 (10%)
Add to cart
Touch Screen Dry Bath/Block Heater
Catalog number88870009
Price (USD)/ Each
1,098.00
Online exclusive
1220.00 
Save 122.00 (10%)
-
Add to cart

Designed to achieve precise temperature stability and uniformity with an easy-to-use touch screen controller. With 1, 2 and 4 block models, a broad range of heat blocks and a wide temperature range of ambient +5°C to 130°C, you're sure to find the dry bath to fit your application demands. The Thermo Scientific™ Touch Screen Dry Bath/Block Heater includes advanced features such as a timer, programmability with the ability to save up to 10 programs with 5 steps per program, and an optional external temperature sensor to control the heat setting by a real time sample read out.

  • Advanced internal temperature sensing probe with outstanding temperature accuracy and control
  • Temperature deviation adjustment
  • Precise temperature control with PID circuit
  • Temperature self-calibration enables user to offset the temperature to desired value
  • Timer enables the user to accurately monitor the heating time
  • Password protection feature
  • Interchangeable aluminum alloy sample blocks designed to provide versatility, easy cleaning and disinfecting
  • Range of configuration to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications
  • Powder-coated steel body construction ensures durability
  • When the external temperature sensor is connected, the internal temperature sensor will disconnect automatically, allowing the temperature control and display to rely on the external temperature sensor
  • Easy-to-use touch screen control displays time and temperature
  • 10 user setting programs, each supports up to 5 sequential steps of different temperatures and duration times
  • Built-in over temperature protection device for added safety
Applications:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation
Required accessory:
  • Unit must be ordered with block(s)
Warranty: 2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE, cULus, RoHS
Controller TypePID digital
LightingLED
MaterialPowder-coated Steel
Plug TypeUS
Relative Humidity<80%
Temperature Range (Metric)Ambient +5° to 130°C (Ambient at 25°C)
TypeDry Bath/Block
Fuse250V 8A
Heating Rate≤ 25 min. 30° to 130°C
IncludesDry bath, US style plug, block lifter
No. of Blocks4 (must be ordered separately)
Temperature Accuracy≤ ± 0.5°C
Temperature Uniformity≤ ± 1°C
Timer0 to 99:59 min or continuous
Warranty2 Year, Replacement
Dimensions (L x W x H)535 x 200 x 100 mm (21.1 x 7.9 x 3.9 in.)
Length (English)21.1 in.
Length (Metric)535 mm
Width (English)7.9 in.
Width (Metric)200 mm
Height (English)3.9 in.
Height (Metric)100 mm
Weight (English)11.24 lb.
Weight (Metric)5.1 kg
Voltage100/120 V
Frequency50/60 Hz
Unit SizeEach