Digital Dry Baths/Block Heaters
Digital Dry Baths/Block Heaters
Thermo Scientific™

Digital Dry Baths/Block Heaters

Increase lab versatility with the Thermo Scientific™ digital dry baths/block heaters.
Have Questions?
Change viewbuttonViewtableView
No. of Blocks:
2 (order separately)
1 (order separately)
4 (order separately)
Heating Rate:
View all
≤ 20 min. 30° to 130°C
Voltage:
100/120 V
200/240 V
Includes:
View all
Dry bath, US style plug, block lifter
Catalog number 88870002
Price (USD)/ Each
331.00
Special Offer
Online exclusive
Ends: 31-Dec-2024
662.00 
Save 331.00 (50%)
-
Add to cart
No. of Blocks:
2 (order separately)
Heating Rate:
≤ 20 min. 30° to 130°C
Voltage:
100/120 V
Includes:
Dry bath, US style plug, block lifter
Price (USD)/ Each
331.00
Special Offer
Online exclusive
Ends: 31-Dec-2024
662.00 
Save 331.00 (50%)
Add to cart
Digital Dry Baths/Block Heaters
Catalog number88870002
Price (USD)/ Each
331.00
Special Offer
Online exclusive
Ends: 31-Dec-2024
662.00 
Save 331.00 (50%)
-
Add to cart
Increase lab versatility with the Thermo Scientific™ digital dry baths, which offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a max temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.
Thermo Scientific digital dry baths are available in a range of configurations to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration allows the user to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device ensures sample and user safety
  • Temperature deviation adjustment
Applications:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation
Required Accessory:
  • Unit must be ordered with block(s)
    Thermo Scientific digital dry baths are available in a range of configurations to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
    • Digital controls and display of time and temperature
    • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
    • Precise temperature control with PID circuit
    • Temperature calibration allows the user to offset the temperature to desired value
    • Timer allows the user to accurately monitor the heating time
    • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
    • Powder-coated steel body construction ensures durability
    • Built-in over-temperature protection device ensures sample and user safety
    • Temperature deviation adjustment
    Applications:
    • Sample preparation
    • Enzyme preservation
    • Enzyme-substrate reactions
    • DNA amplification
    • Electrophoresis gel degeneration
    • Serum coagulation
    Required Accessory:
    • Unit must be ordered with block(s)
Warranty: 2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE, cULus, RoHS
Controller TypePID digital
LightingLED
MaterialPowder-coated Steel
Plug TypeUS
Relative Humidity<80%
Temperature Range (Metric)Ambient +5° to 130°C (Ambient at 25°C)
TypeDry Bath/Block
Fuse250V 5A
Heating Rate≤ 20 min. 30° to 130°C
IncludesDry bath, US style plug, block lifter
No. of Blocks2 (order separately)
Temperature Accuracy≤ ± 0.5°C
Temperature Uniformity≤ ± 1°C
Timer0 to 99:59 min or continuous
Warranty2 Year, Replacement
Dimensions (L x W x H)318 x 200 x 100 mm (12.5 x 7.9 x 3.9 in.)
Length (English)12.5 in.
Length (Metric)318 mm
Width (English)7.9 in.
Width (Metric)200 mm
Height (English)3.9 in.
Height (Metric)100 mm
Weight (English)7.28 lb.
Weight (Metric)3.3 kg
Voltage100/120 V
Frequency50/60 Hz
Unit SizeEach