Slide-A-Lyzer™ MINI Dialysis Devices, 20K MWCO
Slide-A-Lyzer™ MINI Dialysis Devices, 20K MWCO
Thermo Scientific™

Slide-A-Lyzer™ MINI Dialysis Devices, 20K MWCO

Perform sample buffer exchange with these 20,000-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeSample Volume (Metric)Quantity
88405Cup, 50 mL Conical2 mL25 Devices
69590Cup, Microcentrifuge0.1 mL50 Devices
69555Cup, Microcentrifuge0.1 mL250 Devices
88402Cup, 15 mL Conical0.5 mL25 Devices
Catalog number 88405
Price (USD)/ 25 devices
289.65
Online exclusive
313.00 
Save 23.35 (7%)
-
Add to cart
Product Type:
Cup, 50 mL Conical
Sample Volume (Metric):
2 mL
Quantity:
25 Devices
Request bulk or custom format
Price (USD)/ 25 devices
289.65
Online exclusive
313.00 
Save 23.35 (7%)
Add to cart
Slide-A-Lyzer™ MINI Dialysis Devices, 20K MWCO
Catalog number88405
Price (USD)/ 25 devices
289.65
Online exclusive
313.00 
Save 23.35 (7%)
-
Add to cart
Slide-A-Lyzer MINI Dialysis Devices, 20K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules >20 kDa in volumes from 10 μL to 2 mL.
Features of Slide-A-Lyzer MINI Dialysis Devices, 20K MWCO:
Three convenient sizes—dialysis devices for 0.1, 0.5, and 2 mL samples fit standard 1.5-mL micro-centrifuge, 15- and 50-mL conical tubes, respectively
Excellent sample recoveries—low-binding plastic and small membrane surface area minimize sample loss compared to filtration and resin systems
One-step protocol—pipette sample into the MINI device and place in tube containing the dialysis buffer; no laborious assembly, device preparation, or expensive equipment are required
100% leak-tested—innovative design does not permit 'wicking' that can occur in home-made devices
Time savings—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes, which improves sample resolution, quantification, and conjugation

These 20K MWCO dialysis devices are available in three sizes (0.1, 0.5, and 2 mL capacities) and are designed to allow easy sample addition and removal using a standard laboratory pipette. The MINI devices are effective and convenient for accomplishing typical sample desalting or buffer exchange in 4–8 hours. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters, and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with a number of common chemicals and buffers.

Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialysing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

The 0.5- and 2-mL MINI dialysis devices fit in and are capped by inserting them into the included 15- and 50-mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI dialysis devices, low molecular weight contaminant removal and buffer exchange and desalting can be accomplished within 4–8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.

The 0.1 mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL micro-centrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker. Using the 0.1 mL MINI device, 100 μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes.

As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.

For Research Use Only. Not for use in diagnostic procedures.

Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

The 0.5mL and 2mL MINI Dialysis Devices fit in and are capped by inserting them into the included 15mL and 50mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI Dialysis Devices, low molecular weight contaminant removal, buffer exchange and desalting, can be accomplished within 4 to 8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.

The 0.1mL MINI Dialysis Devices are designed to be capped (caps included) and inserted into standard 1.5mL microcentrifuge tubes. Alternatively, as many as 25 MINI Devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple inside a large beaker. Using the 0.1mL MINI Device, 100μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes.

Specifications
Product TypeCup, 50 mL Conical
Product LineSlide-A-Lyzer™
MWCO20 kDa
MembraneCellulose Membrane, Dialysis
FormatMINI Dialysis Device
Purification TargetBuffer Exchange, Protein
Sample Volume (Metric)2 mL
Sufficient For25 Samples, 200–2000 μL each
Quantity25 Devices
Unit Size25 devices
Contents & Storage
Store at room temperature