eBioscience™ Annexin V Apoptosis Detection Kits
Product Image
Invitrogen™

eBioscience™ Annexin V Apoptosis Detection Kits

Kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugateExcitation/EmissionQuantity
88-8008-74PCP-eFluor 710200 Test
88-8005-74FITC, Propidium Iodide488, 535/520, 617200 Tests
88-8006-72eFluor 450, 7-AAD405, 546/450, 64750 Tests
88-8006-74eFluor 450, 7-AAD405, 546/450, 647200 Tests
88-8007-74APC, Propidium Iodide650, 535/660, 617200 Tests
88-8008-72PCP-eFluor 71050 Tests
BMS500FI-100FITC, Propidium Iodide488, 535/520, 617100 Tests
BMS500FI-20FITC, Propidium Iodide488, 535/520, 61720 Tests
BMS500FI-300FITC, Propidium Iodide488, 535/520, 617300 Tests
88-8102-72PE, 7-AAD50 Tests
88-8102-74PE, 7-AAD200 Tests
88-8103-72PE/Cy750 Tests
88-8103-74PE/Cy7200 Tests
BMS500BT-100
also known as BMS500BT/100
Biotin, Propidium IodideNA, 535/NA, 617100 Tests
BMS500BT-20
also known as BMS500BT/20
Biotin, Propidium IodideNA, 535/NA, 61720 Tests
BMS500BT-300
also known as BMS500BT/300
Biotin, Propidium IodideNA, 535/NA, 617300 Tests
88-8005-72FITC, Propidium Iodide488, 535/520, 61750 Tests
88-8007-72APC, Propidium Iodide650, 535/660, 61750 Tests
Catalog number 88-8008-74
Price (USD)/ 200 tests
504.00
-
Add to cart
Conjugate:
PCP-eFluor 710
Quantity:
200 Test
Price (USD)/ 200 tests
504.00
Add to cart
eBioscience™ Annexin V Apoptosis Detection Kits
Catalog number88-8008-74
Price (USD)/ 200 tests
504.00
-
Add to cart
Easily distinguish cells undergoing early and late apoptosis with eBioscience and Annexin V Apoptosis Detection kits for flow cytometry analysis. These kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane. In late-stage apoptosis, as cell integrity is lost, both annexin V and the viability dye traverse and bind PS on the interior of the plasma membrane.
Annexins are a family of calcium-dependent phospholipid-binding proteins that preferentially bind phosphatidylserine (PS). Under normal physiologic conditions, PS is predominantly located in the inner leaflet of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution across the phospholipid bilayer and is translocated to the extracellular membrane leaflet marking cells as targets of phagocytosis. Once on the outer surface of the membrane, PS can be detected by fluorescently labeled annexin V in a calcium-dependent manner.

In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or fixable viability dyes such as eFluor 450. These cells will stain with annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late-stage apoptosis, the cell membrane loses integrity, thereby allowing annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (annexin V, viability dye-positive) from the early-stage apoptotic cells (annexin V positive, viability dye-negative).

Current viability stains available with the eBioscience Annexin V Apoptosis Detection kits include FITC, eFluor 450, allophycocyanin (APC), phycoerythrin (PE), PE-cyanine7, and biotin.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptioneBioscience Annexin V Apoptosis Detection Kit PerCP-eFluor 710
Quantity200 Test
Kit Contents• Annexin V Binding Buffer (10X), 1 bottle (100 mL)
• Annexin V PerCP-eFluor 710, 1 vial (1 mL)

Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits.
Product TypeApoptosis Detection Kit
Shipping ConditionWet Ice
ConjugatePCP-eFluor 710
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Unit Size200 tests
Contents & Storage
2°C to 8°C. Avoid temperature fluctuations.