Catalog Number | Tubing Size |
---|---|
8600-3018 | No. 18 |
8600-3015 | No. 15 |
8600-3016 | No. 16 |
8600-3017 | No. 17 |
8600-3024 | No. 24 |
8600-3025 | No. 25 |
8600-3173 | No. 73 |
8600-3182 | No. 82 |
Ensure reliable and accurate fluid transfers every time with Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing. This flexible, durable high-purity translucent tubing is designed for a variety of transfer applications, including pharmaceutical, laboratory and bioprocess manufacturing.
Ordering Information
Double-bagged in case of 25 in. (7.6m)
Warranty: 90 days
Note: Not intended for invasive use.