Catalog Number | Description |
---|---|
842312052231 | iCAP 7000 Duo High Solids Kit |
842312052251 | iCAP 7000 Duo Volatile Organics Kit |
842312052261 | iCAP 7000 Duo Organics Kit |
842312052271 | iCAP 7000 Radial Aqueous Kit |
842312052301 | iCAP 7000 Radial Volatile Organics Kit |
842312052311 | iCAP 7000 Radial Organics Kit |
842312052291 | iCAP 7000 Radial HF Kit |
842312052281 | iCAP 7000 Radial High Solids Kit |
842312052221 | iCAP 7000 Duo Aqueous Kit |
842312052241 | iCAP 7000 Duo HF Kit |
Aqueous
Supplied as standard with all iCAP 7000 Series ICP-OES models, this aqueous sample introduction kit is ideal for liquid sample solutions containing relatively low acid content (except for hydrofluoric acid (HF)) or up to 3% m/v dissolved solids.
The concentric glass nebulizer within this kit is not suitable for solutions containing large suspended particles due to the risk of blockage.
High Solids
In samples such as sea waters, brines and ground waters, geochemical samples and fusion mixtures, with solid particles in suspension (up to 20% m/v dissolved solids), the high solids kit is necessary to prevent blockage problems. Used in conjunction with an argon humidifier, the kit enables extended analysis times without drift.
Organic
Recommended introduction kit for samples containing relatively nonvolatile, organic solvents, such as xylene, toluene, white spirit and kerosene. These solvents are most likely to be used as diluents for lubricating oils analysis, including additive compounds in virgin oil and wear metals in used oil.
Volatile Organic
Volatile organic solvents, such as alcohols, gasolines and naphthas are often found in petrochemical analysis, both for by-products such as plastics and for petrol itself. The volatile organics sample handling kit is recommended to provide the optimum configuration for analysis by reducing the plasma solvent load.
HF
Un-complexed hydrofluoric acid (HF) is typically obtained during the analysis of geological samples, soils, sediments, ceramics, bricks and cement, together with some types of high temperature alloys. The HF resistant sample handling kit is designed to handle such sample solutions.
Boost Performance with Precision Sample Introduction Kits
Just like our analyzers, our sample introduction kits are designed to maximize convenience, ease of use and precision.
Ordering Requirements
Peristaltic pump tubing must be ordered in addition to the sample introduction kits.