Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Thermo Scientific™

Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers

Simplify sampling with easy access
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)Holds
840-277700Long path rectangular cell holder that accommodates 20-100 mm path length.  Comes assembled in a removable compartment liner.GENESYS 30/40/50 Spectrophotometers20 to 100 mm cuvette (1)
840-298900Standard 10mm single-cell holderGENESYS 140/150/180, BioMate 160 Spectrophotometers10 mm cuvette (1)
840-299000Beckman-style cell holderGENESYS 140/150/180, BioMate 160 SpectrophotometersBeckman-style short microcells
840-303100Long path cylindrical cell holderGENESYS 140/150/180, BioMate 160 Spectrophotometers5 mm to 100 mm cylindrical cuvette (1)
840-303200Sample holder for film/filter accommodates filters/lenses up to 50 mm long, 80mm tall and 10mm thick. Comes assembled in a removable sample compartment liner.GENESYS 140/150/180, BioMate 160 SpectrophotometersFilter/lenses up to 50 mm long x 80 mm tall x 10 mm thick. Minimum filter size 18 mm.
840-303300Disposable microcell holderGENESYS 140/150/180, BioMate 160 SpectrophotometersMicrocells disposable 50 μL plastic Uvettes™
840-303800Long path rectangular cell holderGENESYS 140/150/180, BioMate 160 Spectrophotometers10 mm to 100 mm cuvette (1)
335079-000Thermostatted rectangular cell holderSPECTRONIC 200 and GENESYS 30/40/50 SpectrophotometersSquare 10 mm Pathlength Cuvette
335917-000Adjustable filter holderGENESYS 30/40/50 SpectrophotometersFilters up to 10 mm thick
840-277-500
also known as 840-277500
Test tube and vial holder accommodating 12 mm diameter up to 25 mm diameter test tubes and vialsGENESYS 30/40/50 Spectrophotometers12 mm to 25 mm diameter test tubes and vials
840-277600Test-tube cap covers test-tubes up to 15 cm/6 inches tall to allow measurements while the sample compartment lid is open.GENESYS 30/40/50 SpectrophotometersTall tubes and vials (lid open)
840-277800This sample accessory holds one cylindrical long path cell holder with 5-100 mm path length. Comes assembled in a removable sample compartment liner.GENESYS 30/40/50 Spectrophotometers5 to 100 mm cylindrical cuvette (1)
840-278100Sample holder for film/filter accommodates filters/lenses up to 50 mm long, 80 mm tall and 6 mm thick. Comes assembled in a removable sample compartment liner.GENESYS 30/40/50 SpectrophotometersFilter/lenses up to 50 mm long x 80 mm tall x 6 mm thick
840-278200Accessory Kit for Hose Entry/ExitGENESYS 30/40/50 SpectrophotometersEntry and exit hoses (included)
336028-00010 mm Square Cell HolderGENESYS 30/40/50 SpectrophotometersSingle-cell cuvette or 10 mm diameter test tubes
840-278600Standard 10 mm single-cell holderGENESYS 30/40/50 Spectrophotometers10 mm cuvette (1)
Catalog number 840-277700
Price (USD)/ Each
-
Request A Quote
Description:
Long path rectangular cell holder that accommodates 20-100 mm path length.  Comes assembled in a removable compartment liner.
For Use With (Equipment):
GENESYS 30/40/50 Spectrophotometers
Holds:
20 to 100 mm cuvette (1)

Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.

We provide single-cell solutions for measuring different types of samples.

Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
Specifications
DescriptionLong path rectangular cell holder that accommodates 20-100 mm path length.  Comes assembled in a removable compartment liner.
TypeLong Path Cell Holder
For Use With (Application)Measurements of solutions with low absorbance values
For Use With (Equipment)GENESYS 30/40/50 Spectrophotometers
Holds20 to 100 mm cuvette (1)
ShapeRectangular
Unit SizeEach