Catalog Number | Description | For Use With (Application) |
---|---|---|
839050N | Immunofluorescent assay for quantitative determination of soluble chromogranin A (CgA) | The B·R·A·H·M·S CgA II KRYPTOR test is indicated as an aid to be used in conjunction with clinical evaluation for follow-up and monitoring of patients with neuroendocrine tumors (NETs) and for therapy monitoring of patients with prostate cancer. The test is further indicated as an aid to be used in conjunction with clinical evaluation for the diagnosis of patients with suspected pheochromocytomas. Please consult IFU available in your country. |
083991N | Calibrator for B·R·A·H·M·S CgA II KRYPTOR | Readjust the standard curve memorized in B·R·A·H·M·S KRYPTOR compact PLUS/KRYPTOR GOLD |
083992N | Controls for B·R·A·H·M·S CgA II KRYPTOR (2 levels) | Quality Control for B·R·A·H·M·S CgA II KRYPTOR assay |
Thermo Scientific™ B·R·A·H·M·S™ CgA II KRYPTOR™ is an automated immunofluorescent assay using Time-Resolved amplified Cryptate Emission (TRACE™) technology to determine the concentration of Chromogranin A (CgA) in human serum. B·R·A·H·M·S CgA II KRYPTOR is indicated as an aid to be used in conjunction with clinical evaluation for follow-up and monitoring of patients with neuroendocrine tumors (NETs) and for therapy monitoring of patients with prostate cancer. The test is further indicated as an aid to be used in conjunction with clinical evaluation for the diagnosis of patients with suspected pheochromocytomas. Please consult the Instructions For Use (IFU) available in your country.
Intended Use for US only: B·R·A·H·M·S CgA II KRYPTOR is to be used in conjunction with other clinical methods as an aid in monitoring of disease progression during the course of disease and treatment in patients with gastroentero-pancreatic neuroendocrine tumors (GEP-NETs, grade 1 and grade 2).