Get more precise results than those obtained with conventional electrodes by using the Thermo Scientific™ Orion™ 8162SC ROSS™ Combination Sleeve Junction pH Electrode. This electrode provides stable readings to 0.01 pH in less than 30 seconds.
Includes: