Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes
Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes
Thermo Scientific™

Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes

Measure both pH and temperature in samples with these low maintenance, pH/ATC combination electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesLength (Metric) CableFor Use With (Equipment)
8107UWMMDBNC Waterproof, 8-pin MiniDIN3 mOrion Versa Star, Star A and Dual Star Meters
8107BNUMDBNC Waterproof, 8-pin MiniDIN1 mOrion Versa Star, Star A and Dual Star Meters
8107BNURCABNC Waterproof, RCA1 mMettler Toledo Five and Seven Series pH Meters
Catalog number 8107UWMMD
Price (USD)/ Each
689.00
-
Add to cart
Connector Types:
BNC Waterproof, 8-pin MiniDIN
Length (Metric) Cable:
3 m
For Use With (Equipment):
Orion Versa Star, Star A and Dual Star Meters
Price (USD)/ Each
689.00
Add to cart
Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes
Catalog number8107UWMMD
Price (USD)/ Each
689.00
-
Add to cart

Increase the accuracy of your readings with the Thermo Scientific™ Orion™ ROSS Ultra™ Low Maintenance pH/ATC Triode™ Combination Electrodes. These combination electrodes have a pH sensor that delivers superior performance at varying sample temperatures. They combine the stability, precision and speed of response of the ROSS Ultra line with integrated temperature measurement. Orion ROSS electrodes contain no silver or silver chloride and incorporate a double-junction design, which protects the sample from contamination and makes the electrodes compatible with TRIS buffers.

Measure pH and Temperature with One Low-Maintenance Electrode

Low ionic strength samples, such as clean waters, present a challenge to pH electrode performance. Sluggish response, poor accuracy and drift are common. The Thermo Scientific Orion ROSS Ultra 8107 Triode pH electrode responds within one minute, proving a stable, accurate reading.

Complete your pH measurements faster. Get long, maintenance-free electrode lifetime with special long-life gel and innovative capillary glass junction design. Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.

Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.

Target Markets:

  • Pharmaceutical/Biopharmaceutical
  • Food Manufacturing
  • Academic/Government
  • Environmental

Applications:

  • Quality Control
  • Research
  • General Purpose

Warranty: 18 months from date of purchase by Buyer or 24 months from date of shipment from Seller, whichever is earlier.

Specifications
DescriptionOrion ROSS Ultra Low Maintenance pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 3 meter cable
Connector TypesBNC Waterproof, 8-pin MiniDIN
JunctionGlass Capillary
MaterialRugged Epoxy Body, Filled with Low Maintenance Gel
pH Range0 to 14
Temperature (Metric) Operating0°C to 80°C
TypepH/ATC Combination Electrode
Filling Solution TypeNone required
For Use With (Equipment)Orion Versa Star, Star A and Dual Star Meters
Internal ReferenceROSS
Isopotential Point7
pH Relative Accuracy0.03 pH for buffers using automatic temperature compensation
Precision0.01
Sample CompatibilityTRIS, sulfides, or proteins
Warranty18 Months
Length (Metric)120 mm
Length (Metric) Cable3 m
Diameter (Metric)12 mm
Unit SizeEach