Catalog Number | Capacity (Metric) | Material | Sterility | Packaging |
---|---|---|---|---|
8093-11 | 25 mL | Polystyrene | Sterile | 10 Bags of 10 |
95128095 | 25 mL | Polystyrene | Non-sterile | 10 Bags of 10 |
8096-11 | 25 mL | Plastic | Sterile | Individually Wrapped |
95128085 | 100 mL | Polystyrene | Non-sterile | 10 Bags of 10 |
95128093 | 25 mL | Polystyrene | Non-sterile | Bulk |
Thermo Scientific™ Reagent Reservoirs provide a variety of solutions to suit all of your pipetting procedures.
Innovation goes beyond our pipetters and tips. Even our reagent reservoirs have been designed to provide solutions tailored to your procedures, rather than the other way around. Four variations of reservoirs are offered, 25mL, 25mL with divider, 100mL and a disposable polystyrene 75mL reservoir.
Benefits