Halt™ Phosphatase Inhibitor Single-Use Cocktail
Halt™ Phosphatase Inhibitor Single-Use Cocktail
Thermo Scientific™

Halt™ Phosphatase Inhibitor Single-Use Cocktail

Thermo Scientific Halt Phosphatase Inhibitor Single Use Cocktail preserves the phosphorylation state of proteins during and after cell lysis orRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
784265 x 1 mL
784201 mL
7842824 x 100 μL
7842710 mL
Catalog number 78426
Price (USD)/ 5 x 1 mL
700.65
Online exclusive
778.00 
Save 77.35 (10%)
-
Add to cart
Quantity:
5 x 1 mL
Request bulk or custom format
Price (USD)/ 5 x 1 mL
700.65
Online exclusive
778.00 
Save 77.35 (10%)
Add to cart
Halt™ Phosphatase Inhibitor Single-Use Cocktail
Catalog number78426
Price (USD)/ 5 x 1 mL
700.65
Online exclusive
778.00 
Save 77.35 (10%)
-
Add to cart
Thermo Scientific Halt Phosphatase Inhibitor Single Use Cocktail preserves the phosphorylation state of proteins during and after cell lysis or tissue protein extraction.

Features of Halt Phosphatase Inhibitor Single Use Cocktail:

Devoid of protease inhibitors—To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail.
Dialyzable— Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components.
Supplied as a 100X concentrate—effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X.
Compatibility— compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions.
All-in-one format—inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
Compatible with popular protein assays—works with standard protein assays, including BCA and coomassie (Bradford) assays
Convenient storage—refrigerator (2 to 8°C) storage saves valuable space in the freezer and provides for immediate use

Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases as well as protein tyrosine phosphatases (PTPs) during cell lysis. The cocktail contains a mixture of four inhibitors of broad specificity, including sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract. The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays.

Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell, including signal transduction, cell division and apoptosis. To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis. Halt Phosphatase Inhibitor Cocktail accomplishes this task.

Related Products
Pierce™ Phosphatase Inhibitor Mini Tablets
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypePhosphatase Inhibitor Cocktail
FormLiquid
Product LineHalt™
Reagent TypePhosphatase Inhibitor
InhibitorsPhosphatase Inhibitor
Quantity5 x 1 mL
Unit Size5 x 1 mL
Contents & Storage
Upon receipt store product at 4°C. Do not freeze.