ExoSAP-IT™ PCR Product Cleanup Reagent
ExoSAP-IT™ PCR Product Cleanup Reagent
Applied Biosystems™

ExoSAP-IT™ PCR Product Cleanup Reagent

ExoSAP-IT™ PCR Product Cleanup Reagent is used for enzymatic cleanup of amplified PCR product. It hydrolyzes excess primers and nucleotidesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
78201.1.ML500 Reactions
78205.10.ML5000 Reactions
78202.4X.1.ML2000 Reactions
78200.200.UL100 Reactions
Catalog number 78201.1.ML
Price (USD)/ 500 reactions
542.65
Online exclusive
576.00 
Save 33.35 (6%)
-
Add to cart
No. of Reactions:
500 Reactions
Price (USD)/ 500 reactions
542.65
Online exclusive
576.00 
Save 33.35 (6%)
Add to cart
ExoSAP-IT™ PCR Product Cleanup Reagent
Catalog number78201.1.ML
Price (USD)/ 500 reactions
542.65
Online exclusive
576.00 
Save 33.35 (6%)
-
Add to cart

ExoSAP-IT™ PCR Product Cleanup Reagent is used for enzymatic cleanup of amplified PCR product. It hydrolyzes excess primers and nucleotides in a single step. ExoSAP-IT-purified samples are ready for use in downstream applications such as DNA sequencing or single nucleotide polymorphism (SNP) analysis.

  • Remove excess primers and unincorporated nucleotides enzymatically
  • Prepare DNA sequencing samples with one pipetting step
  • Conserve precious PCR products with up to 100% recovery
  • Minimize column cleanup and sample loss

The ExoSAP-IT one-step solution provides a simple way to enzymatically purify a PCR reaction. The reaction setup is complete with one pipetting step, which is followed by two incubations. The first incubation digests excess primer and dephosphorylates nucleotides. The second, high-temperature incubation inactivates the enzymes.

Enzymatic cleanup is a very efficient method for PCR purification. Adding ExoSAP-IT reagent directly to the PCR product eliminates transfer steps to tubes, wells, or columns and conserves PCR amplicons, helping reduce the chance of cross contamination; no further processing needed.

After enzymatic cleanup with ExoSAP-IT reagent, primers and dNTPs will no longer interfere with DNA sequencing or SNP analysis.

Specifications
DescriptionExoSAP-IT™ PCR Product Clean-up Reagent, 500 Reactions, Recombinant, Dry Ice, DNA Sequencing or Single Nucleotide Polymorphism (SNP) Analysis, Use with PCR Purification
Sample TypePCR Products
For Use With (Application)PCR Purification
No. of Reactions500 Reactions
Product LineExoSAP-IT™
Protein FormRecombinant
Quantity1 mL
Research CategoryGenetic Diseases
Shipping ConditionDry Ice
Unit Size500 reactions
Contents & Storage
Shipped on dry ice. Store at -20°C.