Imject™ Purification Buffer Salts
Imject™ Purification Buffer Salts
Thermo Scientific™
Imject™ Purification Buffer Salts
Thermo Scientific Imject Purification Buffer Salts, formulation: Dry powder for dissolution in 60 mL water to make 0.083M sodium phosphate,Read more
Have Questions?
Catalog number 77159
Price (USD)/ 5 g
59.00
-
Add to cart
Price (USD)/ 5 g
59.00
Add to cart
Imject™ Purification Buffer Salts
Catalog number77159
Price (USD)/ 5 g
59.00
-
Add to cart
Thermo Scientific Imject Purification Buffer Salts, formulation: Dry powder for dissolution in 60 mL water to make 0.083M sodium phosphate, 0.9M sodium chloride, 0.1M sorbitol; pH 7.2. Sufficient for preparing 60 mL of solution for buffer-exchange of immunogens following conjugation.
More Product Data
Carrier protein activation and conjugation data for immunogen preparation
Related Products
Imject™ mcKLH (in PBS)
Imject™ mcKLH Subunits, High Purity Research Grade
Imject™ mcKLH (in MES Buffer)
Imject™ EDC mcKLH Spin Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionImject Purification Buffer Salts
Quantity5 g
Product LineImject™
Unit Size5 g
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.