Specifications
Assay SensitivitySee product documentation
Immunoassay Kit FormatOther Format
Product LinePrioCHECK™, Prionics™
Product TypeELISA Kit
Protein FamilyOther Proteins
Target SpeciesBovine
Gene ID (Entrez)Non-Gene
Gene SymbolNon-Gene
Immunoassay Kit GradeStrip
Target Kit NamedFMDV Type O Antibody
Target Organism ClassFoot and Mouth Disease Virus (FMDV)
Target Species ValidatedAll Sensitive Animals
Unit Size440 samples