Catalog Number | Description | Electrical Requirements |
---|---|---|
75007689 | Sorvall BP 8 Centrifuge | 380/400/415 V ±10% 50 Hz 3-phase |
75007681 | Sorvall BP 8 Centrifuge | 200/208/220/230/240 V ±10% 50/60 Hz, Single Phase |
75007683 | Sorvall BP 16 Centrifuge | 200/208/220/230/240 V ±10% 50/60 Hz, Single Phase |
75007695 | Sorvall BP 16 Centrifuge | 380/400/415 V ±10% 50 Hz 3-phase |
75007662 | Sorvall BP 8 Heavy Duty Water Cooled Centrifuge | 200/208/220/230/240 V ±10% 50/60 Hz, Single Phase |
75007661 | Sorvall BP 16 Heavy Duty Water Cooled Centrifuge | 200/208/220/230/240 V ±10% 50/60 Hz, Single Phase |
75007682 | Sorvall BP 16 Heavy Duty Centrifuge | 200/208/220/230/240 V ±10% 50/60 Hz, Single Phase |
75007684 | Sorvall BP 8 Heavy Duty Centrifuge | 200/208/220/230/240 V ±10% 50/60 Hz, Single Phase |
75007688 | Sorvall BP 16 Heavy Duty Centrifuge | 380/400/415 V ±10% 50 Hz 3-phase |
With an improved capacity of 16 x 500mL blood bags and user-friendly design, the NEW Thermo Scientific™ Sorvall™ BP 8 and 16 blood banking centrifuges make working with large volumes easier and more convenient, using the protocols you've already validated and is in compliance with global standards.
We also offer a range of adapters allowing the centrifugation of most common tubes and microplates, using the same rotor body and oval buckets (75003964).