Nalgene™ Syringe Filters, Non-sterile
Nalgene™ Syringe Filters, Non-sterile
Thermo Scientific™

Nalgene™ Syringe Filters, Non-sterile

Green features
Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDiameter (Metric)Material (Membrane)Unit Size
721-13200.2 μm13 mmPTFECase of 100
199-20200.2 μm25 mmPTFECase of 300
721-13450.45 μm13 mmPTFECase of 100
722-20001.1 μm25 mmGlass FiberCase of 50
724-20200.2 μm25 mmSFCACase of 500
724-2020PK
also known as 724-2020
0.2 μm25 mmSFCAPack of 100
724-20450.45 μm25 mmSFCACase of 500
724-2045PK
also known as 724-2045
0.45 μm25 mmSFCAPack of 100
724-20800.8 μm25 mmCellulose AcetateCase of 500
724-2080PK
also known as 724-2080
0.8 μm25 mmCellulose AcetatePack of 100
727-20200.2 μm25 mmNylonCase of 500
727-2020PK
also known as 727-2020
0.2 μm25 mmNylonPack of 100
727-20450.45 μm25 mmNylonCase of 500
727-2045PK
also known as 727-2045
0.45 μm25 mmNylonPack of 100
728-20200.2 μm25 mmPTFECase of 500
728-2020PK
also known as 728-2020
0.2 μm25 mmPTFEPack of 100
728-20450.45 μm25 mmPTFECase of 500
728-2045PK
also known as 728-2045
0.45 μm25 mmPTFEPack of 100
176-00200.2 μm4 mmNylonCase of 400
176-0020PK
also known as 176-0020
0.2 μm4 mmNylonPack of 100
176-00450.45 μm4 mmNylonCase of 400
176-0045PK
also known as 176-0045
0.45 μm4 mmNylonPack of 100
Catalog number 721-1320
Price (USD)/ Case of 100
311.40
Online exclusive
346.00 
Save 34.60 (10%)
-
Add to cart
Pore Size:
0.2 μm
Diameter (Metric):
13 mm
Material (Membrane):
PTFE
Unit Size:
Case of 100
Price (USD)/ Case of 100
311.40
Online exclusive
346.00 
Save 34.60 (10%)
Add to cart
Nalgene™ Syringe Filters, Non-sterile
Catalog number721-1320
Price (USD)/ Case of 100
311.40
Online exclusive
346.00 
Save 34.60 (10%)
-
Add to cart

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings

Cellulose Acetate (CA)

  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings

Nylon

  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

Specifications
Filter TypeMembrane Filter
Material (Membrane)PTFE
Pore Size0.2 μm
SterilityNon-sterile
For Use With (Application)Sterile venting of air or other gasses, sample preparation of samples for instrument analysis such as HPLC and CG
Housing MaterialPolypropylene (Housing)
Quantity100/Pk.
TypeSyringe Filter
Volume (Metric)2 to 10 mL
Diameter (Metric)13 mm
Unit SizeCase of 100