Use this open access software to customize the interface for individual Chromeleon CDS users for streamlined and easy use.
Use the Thermo Scientific™ Chromeleon™ XPS Open Access software to customize the Thermo Scientific™ Chromeleon™ CDS interface for different users, making it more efficient and easier to use. All users will continue to maintain full capabilities of the CDS software, like instrument control, data processing, reporting, and compliance tools. Chromeleon XPS Open Access enables users to simply walk up and use a very straightforward interface to exploit the full power of the CDS.
Chromeleon XPS Open Access software lets any user run samples on their own by delivering a clean, streamlined walk-up interface that utilizes the full power of Chromeleon CDS. Users can access all instruments controlled by Chromeleon software, including MS, and automatically receive results or notifications about run completion or failure.
Chromeleon XPS software provides error-free execution of any chromatography or MS analysis with minimal training requirements for users.