NanoDrop™ Lite and Lite Plus Printer Paper
NanoDrop™ Lite and Lite Plus Printer Paper
Thermo Scientific™
NanoDrop™ Lite and Lite Plus Printer Paper
Printer paper for the Thermo Scientific™ NanoDrop™ Lite and NanoDrop™ Lite Plus printers.
Have Questions?
Catalog number 714-071400
Price (USD)/ Each
476.00
-
Add to cart
Price (USD)/ Each
476.00
Add to cart
NanoDrop™ Lite and Lite Plus Printer Paper
Catalog number714-071400
Price (USD)/ Each
476.00
-
Add to cart
Printer paper for the Thermo Scientific™ NanoDrop™ Lite and NanoDrop™ Lite Plus printers.
Cryogenic printing paper for use with the NanoDrop Lite and and NanoDrop Lite Plus printer accessories. These labels may be placed in laboratory notebooks or wrapped around microcentifuge tubes being stored in a freezer.
Specifications
DescriptionPrinter Paper
For Use With (Equipment)NanoDrop Lite Spectrophotometer
Unit SizeEach