Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
Thermo Scientific™

Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures

Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.
Have Questions?
Change viewbuttonViewtableView
Product Type:
Replacement Cap
Screw Cap
Stem and Draw Tube
Spout Cap
Closure Size:
38 mm
20 mm
24 mm
28 mm
43 mm
53 mm
63 mm
Color:
View all
Natural
White
Catalog number 712160-0384
Price (USD)/ Case of 12
29.36
Online exclusive
36.70 
Save 7.34 (20%)
-
Add to cart
Product Type:
Replacement Cap
Closure Size:
38 mm
Color:
Natural
Price (USD)/ Case of 12
29.36
Online exclusive
36.70 
Save 7.34 (20%)
Add to cart
Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
Catalog number712160-0384
Price (USD)/ Case of 12
29.36
Online exclusive
36.70 
Save 7.34 (20%)
-
Add to cart

Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.

  • Made to fit Nalgene narrow-mouth and wide-mouth bottles
  • Leakproof† when used with the Nalgene bottles with the matching finish

.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
ColorNatural
DescriptionBottle Replacement Closures
MaterialPolypropylene
No. per Case12
Closure IncludedYes
MouthNarrow
ShapeRound
Product TypeReplacement Cap
For Use With (Equipment)Nalgene narrow-mouth and wide-mouth bottles
Closure MaterialPolypropylene
Closure Size38 mm
Unit SizeCase of 12