Nalgene™ Carboy Replacement Screw Closures and Gaskets
Nalgene™ Carboy Replacement Screw Closures and Gaskets
Actual product may vary
Thermo Scientific™
Nalgene™ Carboy Replacement Screw Closures and Gaskets
Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service
Have Questions?
Change viewbuttonViewtableView
Closure Size:
120 mm
100 mm
53B
83B
Product Type:
View all
Screw Cap
Material:
View all
Polypropylene
Catalog number 712155-1200
Price (USD)/ Case of 12
78.80
Online exclusive
98.50 
Save 19.70 (20%)
-
Add to cart
Closure Size:
120 mm
Product Type:
Screw Cap
Material:
Polypropylene
Price (USD)/ Case of 12
78.80
Online exclusive
98.50 
Save 19.70 (20%)
Add to cart
Nalgene™ Carboy Replacement Screw Closures and Gaskets
Catalog number712155-1200
Price (USD)/ Case of 12
78.80
Online exclusive
98.50 
Save 19.70 (20%)
-
Add to cart
Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service
  • Made to fit Nalgene Carboys
  • Leakproof† when used with the Nalgene bottles with the matching finish
  • † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
    Specifications
    ColorWhite
    DescriptionReplacement Closure
    MaterialPolypropylene
    No. per Case12
    No. per Pack12
    Closure IncludedYes
    Product TypeScrew Cap
    For Use With (Equipment)Nalgene Large Carboys and Bottles
    Closure MaterialPolypropylene
    Closure Size120 mm
    Unit SizeCase of 12