Recover organisms from all patient scenarios, including pediatrics, with just two bottles, reducing inventory and costs and enhancing fastidious organism detection.
Have Questions?
Change view
Catalog Number
Description
Packaging Type
Media Type
Quantity
7102-44
VersaTREK REDOX 1 Media
80 mL/Bottle
Aerobic media
80 mL
7103-44
VersaTREK REDOX 2 Media
80 mL/Bottle
Anaerobic media
80 mL
2 Options
Catalog number 7102-44
Price (USD)/ Each
-
Request A Quote
Description:
VersaTREK REDOX 1 Media
Packaging Type:
80 mL/Bottle
Media Type:
Aerobic media
Quantity:
80 mL
Just two bottles are all you need to recover organisms from all patient scenarios, reducing inventory and costs, with Thermo Scientific™ VersaTREK™ REDOX™ Media. There's no need for costly specialty media, as REDOX 1 aerobic media and REDOX 2 anaerobic media use dilution and timing to detect microbes and are the only media FDA-cleared for draws as low as 0.1mL, perfect for pediatric patients. Plus, the highly-enriched REDOX media supports a wide range of organisms, enhancing the VersaTREK System's ability to better recover fastidious organisms.
Meet the demands of both high and low volume laboratories with the Thermo Scientific VersaTREK System, which comes with a full line of ancillary and accessory products. With four FDA-cleared tests and a simple, two-bottle media system, the VersaTREK System provides maximum value with faster results and fewer limitations.
Largest blood-to-broth dilution ratio in the industry (1:9) to dilute out the effects of antimicrobials, serum host factors and other agents
Enhanced, enriched media to support the wide range of organisms found in the microbiology laboratory
Available in 40 and 80mL sizes and suitable for all patient populations
Only system capable of detecting any gas produced or consumed by organisms, allowing detection of a wider range of both common and fastidious organisms
Simplified workflow includes easy scan function for bottle entry, intuitive icons to reduce training time, and no daily QC requirement for maximum efficiency
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.