Nalgene™ Graduated HDPE Bucket
Nalgene™ Graduated HDPE Bucket
Thermo Scientific™

Nalgene™ Graduated HDPE Bucket

Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle.
Have Questions?
Catalog number 7002-0025
Price (USD)/ Case of 6
621.00
Online exclusive
690.00 
Save 69.00 (10%)
-
Add to cart
Capacity (Metric):
9.5 L
Capacity (English):
2.5 gal.
Price (USD)/ Case of 6
621.00
Online exclusive
690.00 
Save 69.00 (10%)
Add to cart
Nalgene™ Graduated HDPE Bucket
Catalog number7002-0025
Price (USD)/ Case of 6
621.00
Online exclusive
690.00 
Save 69.00 (10%)
-
Add to cart

Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle. This heavy-duty, rugged high density polyethylene bucket is chemically compatible with a wide range of lab reagents.

  • High-density polyethylene construction offers more durability than hard rubber
  • Polyethylene-coated wire bail handle provides a comfortable and sturdy grip
  • Molded-in graduations in pints and liters are easy to read
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 

 

 

Specifications
Capacity (Metric)9.5 L
GraduationsMolded
MaterialHDPE
Capacity (English)2.5 gal.
Description2.5 gal. (9.5 L) Bucket
Handle MaterialPolyethylene Coated
IncludesHandle
Quantity1 Ea.
ShapeWide-Lipped Pour Spout
Unit SizeCase of 6