Slide-A-Lyzer™ Dialysis Cassettes, 7K MWCO
Slide-A-Lyzer™ Dialysis Cassettes, 7K MWCO
Thermo Scientific™

Slide-A-Lyzer™ Dialysis Cassettes, 7K MWCO

Remove salts and contaminants and maximize sample recovery using this semi-permeable membrane with a 7,000-Dalton protein MW cut-off (7K MWCO).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionSample Volume (Metric)
66373Dialysis Cassettes0.5 mL
66370Dialysis Cassettes3 mL
66372Dialysis Cassette Kit3 mL
66710Dialysis Cassettes12 mL
66707Dialysis Cassette Kit12 mL
Catalog number 66373
Price (USD)/ 10 cassettes
114.65
Online exclusive
125.00 
Save 10.35 (8%)
-
Add to cart
Description:
Dialysis Cassettes
Sample Volume (Metric):
0.5 mL
Request bulk or custom format
Price (USD)/ 10 cassettes
114.65
Online exclusive
125.00 
Save 10.35 (8%)
Add to cart
Slide-A-Lyzer™ Dialysis Cassettes, 7K MWCO
Catalog number66373
Price (USD)/ 10 cassettes
114.65
Online exclusive
125.00 
Save 10.35 (8%)
-
Add to cart
Slide-A-Lyzer Dialysis Cassettes (7K MWCO) facilitate the simple and effective removal of salts and small contaminants from proteins or other macromolecules greater than 7,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 12 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange, and more complete sample recovery.
Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
Fast dialysis—flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
Four cassette sizes—select the cassette that best suits your sample volume
Color-coded—green cassette frame specifies cassettes with the 7,000 MW pore size membrane

Membrane specifications:
Composition: regenerated cellulose synthesized by the viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: 13%
Sulfur content: 0.1 to 0.15%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL

Slide-A-Lyzer Dialysis Cassette Technology is protected by U.S. Patent 5,503,741.
Specifications
DescriptionDialysis Cassettes
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO7.0 kDa
Product LineSlide-A-Lyzer™
Quantity10 Cassettes
Sample Volume (Metric)0.5 mL
Unit Size10 cassettes
Contents & Storage
Store at room temperature