Dynabeads™ Streptavidin for Target Enrichment
Dynabeads™ Streptavidin for Target Enrichment
Invitrogen™

Dynabeads™ Streptavidin for Target Enrichment

Dynabeads Streptavidin for Target Enrichment beads are 1-μm monosized, monodispersed, superparamagnetic beads covalently coupled to streptavidin and intended for enrichmentRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
65606D10 mL
65605D2 mL
65607D50 mL
Catalog number 65606D
Price (USD)/ 10 mL
1,413.45
Special Offer
Online exclusive
Ends: 16-Dec-2024
1745.00 
Save 331.55 (19%)
-
Add to cart
Volume (Metric):
10 mL
Price (USD)/ 10 mL
1,413.45
Special Offer
Online exclusive
Ends: 16-Dec-2024
1745.00 
Save 331.55 (19%)
Add to cart
Dynabeads™ Streptavidin for Target Enrichment
Catalog number65606D
Price (USD)/ 10 mL
1,413.45
Special Offer
Online exclusive
Ends: 16-Dec-2024
1745.00 
Save 331.55 (19%)
-
Add to cart

Dynabeads Streptavidin for Target Enrichment beads are 1-μm monosized, monodispersed, superparamagnetic beads covalently coupled to streptavidin and intended for enrichment of target sequences for library preparations for next-generation sequencing (NGS) workflows.

Benefits of Dynabeads Streptavidin for Target Enrichment beads include:
Performance—high sensitivity and reproducibility of results
Efficiency—fast binding kinetics and high binding capacity
Accuracy—excellent coverage, which enables high confidence in variant detection
Automation ready—easy automation for high-throughput needs on the KingFisher Purification System

Consistency and reproducibility  
The monolayer of streptavidin leaves most of the biotin-binding sites available for the binding of free biotin and biotinylated ligands. The specific and defined surface allows for efficient capture, separation, and downstream handling. In addition, the low size distribution (<5%) of the monosized beads and the streptavidin monolayer help ensure negligible leakage, batch consistency, and reproducibility of results.

High binding capacity 
The size of the molecule and the biotinylation procedure will affect the binding capacity. The capacity also depends on steric availability and charge interaction between bead and molecule and between molecules. With two or three biotin binding sites available for each streptavidin molecule on the surface of the bead after immobilization, one mg of Dynabeads Streptavidin for Target Enrichment typically binds: 
• 950–1,500 pmol free biotin 
• ∼20 μg biotinylated ds-DNA 
• ∼400 pmol biotinylated ss-oligonucleotides

Applications
Thermo Fisher streptavidin-coupled Dynabeads magnetic beads have been used and cited for a wide variety of applications. Although the beads can be used to isolate any biotinylated targets, these hydrophobic beads are particularly well suited for the enrichment of target sequences for library preparations for NGS workflows. Dynabeads Streptavidin for Target Enrichment is a high-quality product that enables robust and reproducible NGS analysis results on any NGS platform.

Dynabeads Streptavidin for Target Enrichment efficiently captures DNA sequences of interest through specific hybridization via immobilization of target-specific biotinylated probes to the streptavidin on the bead surface. The subsequent washing steps utilize magnetic separation to obtain high purity. The starting sample can be nucleic acid extracted from chromatin immunoprecipitated samples (ChIP) or isolated pure nucleic acid.

Learn more about our streptavidin products

Automation ready with the KingFisher Purification System
The 1-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher Sample Purification Systems or liquid handlers.

Commercial supply 
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads Streptavidin for Target Enrichment beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or see our Dynabeads OEM page.

For library construction within NGS workflows, Dynabeads Streptavidin for Target Enrichment can be used in conjunction with MagMAX Pure Bind Beads for high-performance library cleanup downstream of your target enrichment protocol.
 

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceISO9001 and ISO13485
Concentration10 mg/mL
ColorBrown
For Use With (Application)Target Sequence Enrichment
MaterialMagnetic Polystyrene
Product TypeStreptavidin Magnetic Beads
For Use With (Equipment)KingFisher™ Sample Purification System, DynaMag™ magnets
High-throughput CompatibilityHigh-throughput Compatible
Isolation TechnologyMagnetic beads
Ligand TypeRecombinant streptavidin
Product LineDynabeads™
Shelf Life48 months from date of manufacture
Shipping ConditionAmbient Temperature
UniformityMonosized 1 μm (CV <5%)
Volume (Metric)10 mL
Diameter (Metric)1 μm
Regulatory StatusFor Research Use Only
Unit Size10 mL
Contents & Storage
Dynabeads Streptavidin for Target Enrichment is supplied in PBS, pH 7.4, with 0.1% BSA, 0.02% NaN3. Store at 2°C to 8°C.