eBioscience™ Fixable Viability Dye eFluor™ 450
Product Image
Invitrogen™

eBioscience™ Fixable Viability Dye eFluor™ 450

Fixable Viability Dye eFluor™ 450 is a viability dye that can be used to irreversibly label dead cells prior toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
65-0863-18500 Tests
65-0863-14100 Tests
Catalog number 65-0863-18
Price (USD)/ 500 tests
359.65
Online exclusive
384.00 
Save 24.35 (6%)
-
Add to cart
Quantity:
500 Tests
Price (USD)/ 500 tests
359.65
Online exclusive
384.00 
Save 24.35 (6%)
Add to cart
eBioscience™ Fixable Viability Dye eFluor™ 450
Catalog number65-0863-18
Price (USD)/ 500 tests
359.65
Online exclusive
384.00 
Save 24.35 (6%)
-
Add to cart
Fixable Viability Dye eFluor™ 450 is a viability dye that can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and propidium iodide, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized, and stained for intracellular antigens without any loss of staining intensity of the dead cells. Thus, using Fixable Viability Dyes allows dead cells to be excluded from analysis when intracellular targets are being studied. Fixable Viability Dyes may be used to label cells from all species.

Fixable Viability Dye eFluor™ 450 can be excited by the violet (405 nm) laser line and has a peak excitation of 450 nm that can be detected using a 450/50 band pass filter (equivalent to eFluor™ 450 or Pacific Blue™). Please make sure that your instrument is capable of detecting this dye. For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye. If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute. After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye.

Fixable Viability Dye eFluor™ 450 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO. It should be protected from light and moisture. Store at -70°C with dessicant. It may be freeze-thawed up to 20 times. Allow vial to equilibrate to room temperature before opening.

Reported Application
Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorBlue
Detection MethodFluorescence
For Use With (Application)Viability Assay
For Use With (Equipment)Flow Cytometer
Product TypeDye
Excitation Wavelength Range405/450
Product LineeBioscience™
Quantity500 Tests
Unit Size500 tests