Nalgene™ Biohazardous Waste Containers
Nalgene™ Biohazardous Waste Containers
Thermo Scientific™

Nalgene™ Biohazardous Waste Containers

Conduct secondary containment of biohazardous waste with these biohazardous waste containers, which feature convenient one-step sterilization and enable one-handed disposal of materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) OuterHeight (Metric)
6370-00045.5 L22 cm28 cm
6370-000519 L28 cm43 cm
6370-001557 L34 cm74 cm
Catalog number 6370-0004
Price (USD)/ Each
192.60
Online exclusive
214.00 
Save 21.40 (10%)
-
Add to cart
Capacity (Metric):
5.5 L
Diameter (Metric) Outer:
22 cm
Height (Metric):
28 cm
Price (USD)/ Each
192.60
Online exclusive
214.00 
Save 21.40 (10%)
Add to cart
Nalgene™ Biohazardous Waste Containers
Catalog number6370-0004
Price (USD)/ Each
192.60
Online exclusive
214.00 
Save 21.40 (10%)
-
Add to cart

Conduct secondary containment of biohazardous waste with Thermo Scientific™ Nalgene™ Biohazardous Waste Containers, which feature convenient one-step sterilization—no need to remove the bag. A port opening in the cover allows for one-handed disposal of materials, limiting exposure to infectious waste. The containers hold standard sized autoclave bags (not included).

  • Polypropylene copolymer (PPCO) components will not rust, dent or puncture like other materials
  • Complies with U.S. OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
  • Autoclavable/Biohazard
  • Symbol printed in English and Spanish
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
 

.
Specifications
Capacity (English)1.5 gal.
Certifications/ComplianceU.S. OSHA Standard 29 CFR Part 1910.1030
Diameter (English) Outer8.75 in.
Capacity (Metric)5.5 L
MaterialPolypropylene
SterilityNon-sterile
TypeWaste Container Biohazardous
Height (English)11 in.
Height (Metric)28 cm
No. per Case1
No. per Pack1
Diameter (Metric) Outer22 cm
Unit SizeEach