Nalgene™ Tubing Y-Type Connectors
Nalgene™ Tubing Y-Type Connectors
Thermo Scientific™
Nalgene™ Tubing Y-Type Connectors
Maximize liquid flow with non-tapered hose barbed Y-Type Connectors, molded of polypropylene resin, free of animal derived components.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
6152-01251/8 in. I.D.
6152-0125PK1/8 in. I.D.
126152-01251/8 in. Tubing
6152-01873/16 in. I.D.
6152-0187PK3/16 in. I.D.
6152-02501/4 in. I.D.
6152-0250PK1/4 in. I.D.
6152-03125/16 in. I.D.
6152-0312PK5/16 in. I.D.
6152-03753/8 in.
6152-0375PK3/8 in.
6152-05001/2 in. Tubing
6152-0500PK1/2 in. Tubing
Catalog number 6152-0125
Price (USD)/ Case of 72
302.00
-
Add to cart
For Use With:
1/8 in. I.D.
Price (USD)/ Case of 72
302.00
Add to cart
Nalgene™ Tubing Y-Type Connectors
Catalog number6152-0125
Price (USD)/ Case of 72
302.00
-
Add to cart
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
  • Polypropylene construction
  • Autoclavable
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Product TypeTubing Connector
DescriptionY-Type Connector
AutoclavableAutoclavable
MaterialPolypropylene
Diameter (English) Outer1/8 in.
Diameter (Metric) Outer3.175 mm
For Use With1/8 in. I.D.
Packaging Quantity12/Pk., 72/Cs.
Unit SizeCase of 72