UltiMate™ 3000 Variable Wavelength Detector Flow Cells
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Actual product may vary
Thermo Scientific™
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Choose the Thermo Scientific™ Dionex™ UltiMate™ 3000 Variable Wavelength Detector Flow Cell based on the flow rate you need.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)TypeMaterial
6074.0200UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Analytical Flow CellPEEK
6074.0320Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Semi-Preparative Flow CellPEEK
6074.0250UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Analytical Flow CellStainless Steel
6074.0270UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Nano Flow CellFused Silica
6074.0280UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Capillary Flow CellFused Silica
6074.0285Dionex UltiMate 3000 Variable Wavelength DetectorUltra-Low Dispersion UV-MonitorFused Silica
6074.0290Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Mic Flow CellFused Silica
6074.0300UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Semi-Micro Flow CellPEEK
6074.0360
also known as 6074.036
UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Semi-Micro Flow CellStainless Steel
Catalog number 6074.0200
Price (USD)/ Each
-
Request A Quote
For Use With (Equipment):
UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Type:
Analytical Flow Cell
Material:
PEEK
Choose the Thermo Scientific™ Dionex™ UltiMate™ 3000 Variable Wavelength Detector Flow Cell based on the flow rate you need. They are available in a variety of diameters for analytical, semi-analytical, semi-preparative and semi-micro flow rates.
Specifications
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialPEEK
Product LineUltiMate
TypeAnalytical Flow Cell
Volume (Metric)11 μL
Diameter (Metric) Outer10 mm
Unit SizeEach