SOLA™ SPE Plates
SOLA™ SPE Plates
Actual product may vary
Thermo Scientific™
SOLA™ SPE Plates
Innovative Thermo Scientific™ SOLA™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionSorbent
60509-003SOLA SAX SPE 30 mg/2 mL 96-well plateMixed Mode Strong Anion Exchanger, Polymeric
60309-00110 mg/2 mL; SOLA 96-well SPE plate; 1 Pk.HRP (Reversed Phase Polymeric)
60509-001SOLA HRP SPE 30 mg/2 mL 96-well plateHRP (Reversed Phase Polymeric)
60309-00210 mg/2 mL; SOLA-CX 96 well SPE plate; 1 Pk.Mixed Mode Strong Cation Exchanger, Polymeric
60509-002SOLA SCX SPE 30 mg/2 mL 96-well plateMixed Mode Strong Cation Exchanger, Polymeric
60309-00310 mg/2 mL; SOLA-AX 96 well SPE plate; 1 Pk.Mixed Mode Strong Anion Exchanger, Polymeric
60309-00410 mg/2 mL; SOLA WCX 96 well SPE plate; 1 Pk.Mixed Mode Weak Cation Exchanger, Polymeric
60509-004SOLA WCX SPE 30 mg/2 mL 96-well plateMixed Mode Weak Cation Exchanger, Polymeric
60309-00510 mg/2 mL; SOLA WAX 96 well SPE plate; 1 Pk.Mixed Mode Weak Anion Exchanger, Polymeric
60509-005SOLA WAX SPE 30 mg/2 mL 96-well plateMixed Mode Weak Anion Exchanger, Polymeric
Catalog number 60509-003
Price (USD)/ Each
362.03
-
Add to cart
Description:
SOLA SAX SPE 30 mg/2 mL 96-well plate
Sorbent:
Mixed Mode Strong Anion Exchanger, Polymeric
Price (USD)/ Each
362.03
Add to cart
SOLA™ SPE Plates
Catalog number60509-003
Price (USD)/ Each
362.03
-
Add to cart
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, SOLA SPE Plates deliver unparalleled performance, including:
  • Higher levels of reproducibility
  • Lower elution volumes
  • High extract cleanliness
SOLA SPE Plates achieve excellent recovery levels even with low volumes of extract solvents, resulting in a more concentrated analyte and increased sensitivity. You’ll also realize additional cost and time saving from faster sample dry-down time and reduced solvent usage.
  • 10mg/2mL 96 well plate format
  • Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
  • SOLA products are also available in cartridge format.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSOLA SAX SPE 30 mg/2 mL 96-well plate
Bed Weight30 mg
For Use With (Application)Bioanalytical Applications - Strong Ion-Exchange Retention of Weak Acidic Compounds. Complementary Reversed Phase Retention of Neutral Compounds.
Product LineSOLA
SorbentMixed Mode Strong Anion Exchanger, Polymeric
Type96-well SPE Plate
Volume (Metric)2 mL
Unit SizeEach