SOLA™ SPE Cartridges
SOLA™ SPE Cartridges
Actual product may vary
Thermo Scientific™
SOLA™ SPE Cartridges
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionSorbent
60409-005SOLA WAX SPE Cartridge 30 mg/3 mLMixed Mode Weak Anion Exchanger, Polymeric
60109-001SOLA HRP SPE CartridgeHRP (Reversed Phase Polymeric)
60409-001SOLA HRP SPE Cartridge 30 mg/3 mLHRP (Reversed Phase Polymeric)
60109-002SOLA SCX SPE CartridgeMixed Mode Strong Cation Exchanger, Polymeric
60409-002SOLA SCX SPE Cartridge 30 mg/3 mLMixed Mode Strong Cation Exchanger, Polymeric
60109-003SOLA SAX SPE CartridgeMixed Mode Strong Anion Exchanger, Polymeric
60409-003SOLA SAX SPE Cartridge 30 mg/3 mLMixed Mode Strong Anion Exchanger, Polymeric
60109-004SOLA WCX SPE CartridgeMixed Mode Weak Cation Exchanger, Polymeric
60409-004SOLA WCX SPE Cartridge 30 mg/3 mLMixed Mode Weak Cation Exchanger, Polymeric
60109-005SOLA WAX SPE CartridgeMixed Mode Weak Anion Exchanger, Polymeric
Catalog number 60409-005
Price (USD)/ Pack of 50
185.74
-
Add to cart
Description:
SOLA WAX SPE Cartridge 30 mg/3 mL
Sorbent:
Mixed Mode Weak Anion Exchanger, Polymeric
Price (USD)/ Pack of 50
185.74
Add to cart
SOLA™ SPE Cartridges
Catalog number60409-005
Price (USD)/ Pack of 50
185.74
-
Add to cart
Grow your laboratory productivity. Thermo Scientific™ SOLA™ SPE Cartridges revolutionize solid phase extraction (SPE). This first fritless SPE product range enables greater sample throughput in bioanalytical laboratories where low elution volumes combined with greater reproducibility, lower failure rates and cleaner extracts are key to achieving better productivity.
SOLA products are available in a 10mg/1mL cartridge format. They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of SOLA cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
SOLA cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What SOLA Cartridges Mean to Your Analysis
  • Higher levels of reproducibility
  • Higher levels of extract cleanliness
  • Reduced solvent requirements
  • Confidence in results
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSOLA WAX SPE Cartridge 30 mg/3 mL
Bed Weight30 mg
For Use With (Application)Bioanalytical Applications - Strong Ion-Exchange Retention of Strong Acidic Compounds. Sorbent Charge Can be Activated or Deactivated. Complementary Reversed Phase Retention of Neutral Compounds.
Product LineSOLA
SorbentMixed Mode Weak Anion Exchanger, Polymeric
TypeSPE Cartridge
Volume (Metric)3 mL
Unit SizePack of 50